Merck
[Syphilis].
Presse medicale (Paris, France : 1983) (2013-02-19)
Nicolas Dupin, David Farhi
ABSTRACT
Syphilis is back since 2000. Early syphilis comprises primary syphilis, secondary syphilis and early latent syphilis (less than 1 year duration). During early phases of syphilis, patients are more contagious and neurologic complications are rare. Early neurosyphilis are mostly represented by uveitis or cranial nerves lesions. Treatment of non-neurologic syphilis are based on intramusculary injection of benzathine-penicilline G: one injection in case of early syphilis, three injections in case of late syphilis. The follow-up after treatment is based on clinical evolution and the titer of VDRL. Intravenously infusion of penicillin G is the only treatment recommended for neurosyphilis.
Sigma-Aldrich
Penicillin G sodium salt, 96.0-102.0%
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Supelco
Penicillin G potassium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Penicillin G potassium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G potassium salt, 95.0 - 102.0%
Sigma-Aldrich
Penicillin G potassium salt