Skip to Content
Merck
All Photos(1)

Documents

46401

Supelco

Cycloheximide

PESTANAL®, analytical standard

Synonym(s):

3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide, Actidione, Naramycin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
Beilstein:
88868
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of stereo isomers

product line

PESTANAL®

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: slightly soluble

antibiotic activity spectrum

fungi

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC2CC(=O)NC(=O)C2

InChI

1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1

InChI key

YPHMISFOHDHNIV-FSZOTQKASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cycloheximide is an antibiotic having significant antifungal properties. It is produced by streptomycin-producing strains of Streptomyces griseus and acts by inhibiting protein synthesis. Its main biological activity is translation inhibition in eukaryotes resulting in cell growth arrest and cell death. It is used as a fungicide.
As per Regulation (EC) No 1107/2009, repealing the directive 91/414, cycloheximide is not approved for use in the European Union (EU).

Application

Cycloheximide may find the following uses:
  • To evaluate the antifungal efficacy and safety of cycloheximide as a supplement in Optisol-GS
  • To study the effect of cycloheximide on leaf-cutting ant workers
  • To study the effect of streptomycin, cycloheximide, fungizone, captan, carbofuran, cygon, and pentachloronitrobenzene on soil microorganisms
  • To study the fungal control of nitrous oxide production in a semiarid grassland
  • To study the activity and degradation of streptomycin and cycloheximide in soil

Biochem/physiol Actions

Mode of Action: Translation inhibition in eukaryotes resulting in cell growth arrest and cell death. CHX is widely used for the selection of CHX-resistant strains of yeast and fungi, controlled inhibition of protein synthesis for detection of short-lived proteins and super-induction of protein expression, and apoptosis induction or facilitation of apoptosis induction by death receptors.

Activity Spectrum: Active against yeast and fungi like Candida, Aspergillus, Saccharomyces, Penicillium

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Muta. 2 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S-H Chan et al.
Oncogene, 33(36), 4496-4507 (2014-03-13)
Metastasis is the predominant cause of death in breast cancer patients. Several lines of evidence have shown that microRNAs (miRs) can have an important role in cancer metastasis. Using isogenic pairs of low and high metastatic lines derived from a
Yuh-Shyan Tsai et al.
Oncotarget, 5(15), 6425-6436 (2014-08-15)
Altered DAB2IP gene expression often detected in prostate cancer (PCa) is due to epigenetic silencing. In this study, we unveil a new mechanism leading to the loss of DAB2IP protein; an oncogenic S-phase kinase-associated protein-2 (Skp2) as E3 ubiquitin ligase
Stephan Meister et al.
Science (New York, N.Y.), 334(6061), 1372-1377 (2011-11-19)
Most malaria drug development focuses on parasite stages detected in red blood cells, even though, to achieve eradication, next-generation drugs active against both erythrocytic and exo-erythrocytic forms would be preferable. We applied a multifactorial approach to a set of >4000
Minako Kaneda et al.
Plant physiology, 147(4), 1750-1760 (2008-06-14)
Secondary xylem (wood) formation in gymnosperms requires that the tracheid protoplasts first build an elaborate secondary cell wall from an array of polysaccharides and then reinforce it with lignin, an amorphous, three-dimensional product of the random radical coupling of monolignols.
Matthias Rottmann et al.
Science (New York, N.Y.), 329(5996), 1175-1180 (2010-09-04)
Recent reports of increased tolerance to artemisinin derivatives--the most recently adopted class of antimalarials--have prompted a need for new treatments. The spirotetrahydro-beta-carbolines, or spiroindolones, are potent drugs that kill the blood stages of Plasmodium falciparum and Plasmodium vivax clinical isolates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service