Skip to Content
Merck
All Photos(1)

Key Documents

46286

Supelco

Flurtamone

PESTANAL®, analytical standard

Synonym(s):

5-(Methylamino)-2-phenyl-4-[3-(trifluoromethyl)phenyl]-3(2H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14F3NO2
CAS Number:
Molecular Weight:
333.30
Beilstein:
7543933
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC1=C(C(=O)C(O1)c2ccccc2)c3cccc(c3)C(F)(F)F

InChI

1S/C18H14F3NO2/c1-22-17-14(12-8-5-9-13(10-12)18(19,20)21)15(23)16(24-17)11-6-3-2-4-7-11/h2-10,16,22H,1H3

InChI key

NYRMIJKDBAQCHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flurtamone is a pre- and post-emergence herbicide, used for the control of broadleaved and grass weeds in soybeans, cotton, peas, groundnuts and sunflower cultivation at pre-plant stage. Its mode of action involves the blocking of carotenoid biosynthesis in weeds via inhibiting the phytoene desaturase.

Application

Flurtamone may be used as an analytical reference standard for the quantification of the analyte in environmental samples using ultra-performance liquid chromatography (UPLC) coupled to tandem mass spectrometry (MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultra-pressure liquid chromatography-electrospray tandem mass spectrometry for multiresidue determination of pesticides in water.
Gervais G, et al.
Journal of Chromatography A, 1202(2), 163-172 (2008)
Paranjape K, et al. et al.
The Pesticide Encyclopedia (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service