
Flurtamone

Synonym(s):
5-(Methylamino)-2-phenyl-4-[3-(trifluoromethyl)phenyl]-3(2H)-furanone
Empirical Formula (Hill Notation):
C18H14F3NO2
CAS Number:
Molecular Weight:
333.30
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2