Skip to Content
Merck
All Photos(1)

Documents

456756

Sigma-Aldrich

Iodomethane solution

2.0 M in tert-butyl methyl ether, contains copper as stabilizer

Synonym(s):

Methyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
Beilstein:
969135
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

16.85 psi ( 55 °C)
4.86 psi ( 20 °C)

Quality Level

contains

copper as stabilizer

concentration

2.0 M in tert-butyl methyl ether

bp

41-43 °C

density

0.933 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CI

InChI

1S/CH3I/c1-2/h1H3

InChI key

INQOMBQAUSQDDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is 2M solution of iodomethane in tert-butyl methyl ether. Iodomethane also known as methyl iodide is an alkyl halide commonly employed as methylating agent.

Application

  • 8-plex LC-MS/MS Analysis of Permethylated N-Glycans Achieved by Using Stable Isotopic Iodomethane.: This research demonstrates the use of iodomethane in stable isotope labeling for the mass spectrometric analysis of permethylated N-glycans. The method enhances the accuracy and sensitivity of glycomic studies (Dong et al., 2019).
  • Comparative glycomic profiling of isotopically permethylated N-glycans by liquid chromatography/electrospray ionization mass spectrometry.: The study presents a comparative analysis of N-glycan profiles using isotopically labeled iodomethane, improving the resolution and identification of glycan structures in complex biological samples (Hu et al., 2013).
  • Antifungal property of quaternized chitosan and its derivatives.: This paper explores the synthesis of quaternized chitosan derivatives using iodomethane and their subsequent antifungal activities. The results suggest potential applications in biomedical and agricultural fields (Sajomsang et al., 2012).
  • Validation of two fluoro-analogues of N,N-dimethyl-2-(2′-amino-4′-hydroxymethyl-phenylthio)benzylamine as serotonin transporter imaging agents using microPET.: This research validates fluoro-analogues of a serotonin transporter imaging agent, synthesized using iodomethane, highlighting its potential in neuroimaging applications (Jarkas et al., 2010).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F

Flash Point(C)

-18 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective mono-methylation of arylacetonitriles and methyl arylacetates by dimethyl carbonate.
Selva M, et al.
Journal of the Chemical Society. Perkin Transactions 1, 10, 1323-1328 (1994)
Xiao-Dan Shi et al.
Food chemistry, 271, 338-344 (2018-09-22)
Dictyophora echinovolvata is a kind of edible mushroom in the Dictyophora genus, of which polysaccharide is an important chemical substance. Herein, three polysaccharide fractions (DEP-4P, DEP-6P and DEP-8P) were prepared from water extract of D. echinovolvata using gradient ethanol precipitation
R Otto et al.
Nature chemistry, 4(7), 534-538 (2012-06-22)
Solvents have a profound influence on chemical reactions in solution and have long been used to control their outcome. Such effects are generally considered to be governed by thermodynamics; however, little is known about the steric effects of solvent molecules.
Nicole J Rijs et al.
Dalton transactions (Cambridge, England : 2003), 39(37), 8655-8662 (2010-08-18)
A combination of multistage mass spectrometry experiments and DFT calculations were used to examine the synthesis and reactivity of dimethylaurate. Collision induced dissociation (CID) of [(CH(3)CO(2))(4)Au](-) proceeded via reductive elimination of acetylperoxide to yield the diacetate [CH(3)CO(2)AuO(2)CCH(3)](-), which in turn
Christian R Evenhuis et al.
The journal of physical chemistry. A, 115(23), 5992-6001 (2011-02-12)
The photodissociation of methyl iodide in the A band is studied by full-dimensional (9D) wave packet dynamics calculations using the multiconfigurational time-dependent Hartree approach. The potential energy surfaces employed are based on the diabatic potentials of Xie et al. [J.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service