![Iodomethane solution 2.0 M in tert-butyl methyl ether, contains copper as stabilizer](/deepweb/assets/sigmaaldrich/product/structures/170/512/9c77d647-4508-4465-bd7f-3de433be487e/640/9c77d647-4508-4465-bd7f-3de433be487e.png)
Iodomethane solution
![Iodomethane solution 2.0 M in tert-butyl methyl ether, contains copper as stabilizer](/deepweb/assets/sigmaaldrich/product/structures/170/512/9c77d647-4508-4465-bd7f-3de433be487e/640/9c77d647-4508-4465-bd7f-3de433be487e.png)
Synonym(s):
Methyl iodide
Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing