Skip to Content
Merck
All Photos(1)

Key Documents

45613

Supelco

Atrazine-desethyl-desisopropyl-2-hydroxy

PESTANAL®, analytical standard

Synonym(s):

4,6-Diamino-2-hydroxy-1,3,5-triazine, Ammeline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N5O
CAS Number:
Molecular Weight:
127.10
Beilstein:
607753
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1nc(N)nc(O)n1

InChI

1S/C3H5N5O/c4-1-6-2(5)8-3(9)7-1/h(H5,4,5,6,7,8,9)

InChI key

MASBWURJQFFLOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongkai Zhu et al.
Journal of hazardous materials, 373, 671-677 (2019-04-08)
Occurrence, spatial distribution, and temporal trends of melamine and its derivatives (ammeline, ammelide, and cyanuric acid) were investigated in surface sediment and two sediment cores collected from Lake Shihwa, South Korea. ∑Melamine (sum of melamine and its three derivatives) was
Hongkai Zhu et al.
Environment international, 123, 375-381 (2018-12-21)
Melamine is used extensively in household products, such as furniture, dinnerware, and food utensils. Several studies have shown that melamine adversely affects kidney function. Nevertheless, little is known about urinary melamine concentrations, and its temporal variability. In this study, 213
Jennifer L Seffernick et al.
Journal of bacteriology, 192(4), 1106-1112 (2009-12-22)
Melamine toxicity in mammals has been attributed to the blockage of kidney tubules by insoluble complexes of melamine with cyanuric acid or uric acid. Bacteria metabolize melamine via three consecutive deamination reactions to generate cyanuric acid. The second deamination reaction
Kazuhiro Takagi et al.
Applied microbiology and biotechnology, 94(6), 1647-1656 (2011-11-23)
Melamine has recently been recognized as a food contaminant with adverse human health effects. Melamine contamination in some crops arises from soil and water pollution from various causes. To remove melamine from the polluted environment, a novel bacterium, Nocardioides sp.
Yanying Li et al.
Environment international, 130, 104924-104924 (2019-07-02)
Traditional extraction methods (soil solution and solvent extraction) are simple to use and conventionally employed to assess pesticide chemical form and bioavailability in soils. However, whilst convenient for regulatory testing, it has been suggested that these approaches may be too

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service