M2659
Melamine
99%
Synonym(s):
2,4,6-Triamino-1,3,5-triazine, sym-Triaminotriazine
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Empirical Formula (Hill Notation):
C3H6N6
CAS Number:
Molecular Weight:
126.12
Beilstein:
124341
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
powder
mp
>300 °C (lit.)
solubility
water: soluble 25 mg/mL, clear to slightly hazy, colorless
SMILES string
Nc1nc(N)nc(N)n1
InChI
1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)
InChI key
JDSHMPZPIAZGSV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Melamine can be used as a:
- Reactant in the preparation of melamine trisulfonic acid (MTSA) which is used as a catalyst in various reactions.
- Nitrogen source for the large scale synthesis of nitrogen-doped graphene (NG).
- Heterogeneous catalyst in combination with lewis acid ZnI2 for conversion of carbon dioxide to cyclic carbonates.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Repr. 2 - STOT RE 2
Target Organs
Urinary tract
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
572.0 °F - closed cup
Flash Point(C)
300 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Melamine trisulfonic acid (MTSA): a new efficient catalyst for the chemoselective methoxymethylation of alcohols.
Shirini F, et al.
Synthetic Communications, 40(6), 910-914 (2010)
Effect of melamine and its salts on combustion and thermal decomposition of polyamide 6.
Levchik SV, et al.
Fire and Materials, 21(2), 75-83 (1997)
Melamine-ZnI2 as heterogeneous catalysts for efficient chemical fixation of carbon dioxide to cyclic carbonates.
Liu M, et al.
Royal Society of Chemistry Advances, 5(2), 960-966 (2015)
The crystal structure of melamine.
Hughes EW.
Journal of the American Chemical Society, 63(6), 1737-1752 (1941)
Infrared and Raman spectra of melaminium chloride hemihydrate.
Marchewka M K.
Materials Science and Engineering, B, 95(3), 214-221 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service