Skip to Content
Merck
All Photos(1)

Key Documents

45466

Supelco

Dodin

PESTANAL®, analytical standard

Synonym(s):

1-Dodecyl-guanidiniumacetat

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H33N3O2
CAS Number:
Molecular Weight:
287.44
Beilstein:
6546960
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(O)=O.CCCCCCCCCCCCNC(N)=N

InChI

1S/C13H29N3.C2H4O2/c1-2-3-4-5-6-7-8-9-10-11-12-16-13(14)15;1-2(3)4/h2-12H2,1H3,(H4,14,15,16);1H3,(H,3,4)

InChI key

YIKWKLYQRFRGPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Cabral
Canadian journal of microbiology, 38(2), 115-123 (1992-02-01)
Treatment of Pseudomonas syringae cells with 50 microM dodecylguanidine monoacetate (dodine) resulted in the rapid degradation and release of RNA and cell lysis. Higher concentrations resulted in a progressive decrease in the intensity of these responses, and the appearance of
R Assini et al.
La Medicina del lavoro, 85(4), 321-326 (1994-07-01)
Current pesticide exposure levels can induce a state of sensitization in exposed workers and only in exceptional situations cause severe intoxication. Three cases are reported--one suffering from urticaria/angio-edema, one from asthma and the other from oculo-rhinitis. Only one subject was
J P Cabral
Journal of basic microbiology, 33(4), 219-225 (1993-01-01)
The release of K+ from Pseudomonas syringae cells treated with dodecylguanidine monoacetate (dodine) was followed with a K(+)-selective glass electrode. Treatment of the cells with 5-15 mumol/l dodine resulted in low levels of K+ release, but higher surfactant concentrations caused
[Determination of dodine in selected plant material].
M Kwast
Roczniki Panstwowego Zakladu Higieny, 32(3), 241-244 (1981-01-01)
M Mitjans et al.
Journal of toxicology and environmental health, 52(6), 545-556 (1997-12-16)
The effect of Dodine on the intestine was studied after a single administration of 1000 mg/kg, which corresponds to the LD50 in male Wistar rats. At this dose, a significant decrease in body weight was observed, accompanied by diarrhea, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service