Skip to Content
Merck
All Photos(1)

Key Documents

D67820

Sigma-Aldrich

2,6-Dichloro-4-nitroaniline

96%

Synonym(s):

Dichloran

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
207.01
Beilstein:
1459581
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

powder

mp

190-192 °C (lit.)

SMILES string

Nc1c(Cl)cc(cc1Cl)[N+]([O-])=O

InChI

1S/C6H4Cl2N2O2/c7-4-1-3(10(11)12)2-5(8)6(4)9/h1-2H,9H2

InChI key

BIXZHMJUSMUDOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Käfer
Mutation research, 241(1), 49-66 (1990-05-01)
Both bleomycin, an antineoplastic drug, and botran (2,6-dichloro-4-nitro-aniline), a fungicide, are known to inhibit growth and induce genetic segregation in diploid tester strains of Aspergillus nidulans when present in agar media. To identify primary effects, samples of induced apparent crossover
L A Myers et al.
Toxicology and applied pharmacology, 95(1), 139-152 (1988-08-01)
The metabolism of 2,6-dichloro-4-nitroaniline (DCNA) to a unique denitrosated product, 3,5-dichloro-p-aminophenol (DCAP), was investigated in rat hepatic microsomes using an HPLC system containing a reverse-phase column and an electrochemical detector. The parent compound appears to induce its own metabolism. The
Ryota Uehara et al.
Cell motility and the cytoskeleton, 65(2), 100-115 (2007-10-31)
Phosphorylation of myosin regulatory light chain (RLC) at Ser19 (mono-phosphorylation) promotes filament assembly and enhances actin-activated ATPase activity of non-muscle myosin, while phosphorylation at both Ser19 and Thr18 (di-phosphorylation) further enhances the ATPase activity. However, it has not well been
H A Hasan
Folia microbiologica, 38(4), 295-298 (1993-01-01)
The effect of fungicides on the production of aflatoxin by Aspergillus flavus IMI 89717, diacetoxyscirpenol and zearalenone by Fusarium graminearum was studied. In a yeast extract-sucrose medium, dicloran, iprodione and vinclozolin fungicides significantly inhibited mycelial growth of A. flavus at
Danielle Palma de Oliveira et al.
Environmental toxicology and chemistry, 28(9), 1881-1884 (2009-05-05)
2,6-Dichloro-4-nitroaniline (dicloran) is a mutagenic aromatic amine used as an agricultural fungicide and in the synthesis of disperse dyes. It is a known mutagen (Salmonella/microsome assay) in strains TA98 and TA100. Dicloran was initially detected, but not quantified, in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service