Skip to Content
Merck
All Photos(1)

Key Documents

43809

Supelco

Perfluorohexanoic acid

analytical standard

Synonym(s):

Undecafluorohexanoic acid, PFAS, Perfluorocaproic acid, Perfluorohexanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)4COOH
CAS Number:
Molecular Weight:
314.05
Beilstein:
1805852
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for ISO 21675 2019

reg. compliance

suitable for FDA C-010.02

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

density

1.759 g/mL at 20 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C6HF11O2/c7-2(8,1(18)19)3(9,10)4(11,12)5(13,14)6(15,16)17/h(H,18,19)

InChI key

PXUULQAPEKKVAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Perfluorohexanoic acid belonging to the class of perfluorinated carboxylic acids (PFCAs), with a 5-carbon perfluoroalkyl chain, was developed as an alternative to the environmental pollutant, perfluorooctanoic acid (PFOA).[1] The degradation of perfluorohexanoic acid is complex, due to the stability of the carbon-fluorine (C-F) bond.[2] It is most commonly used as a wetting agent, corrosion inhibitor, lubricant, and foam fire extinguishant.[3][4]

Application

Perfluorohexanoic acid may be used as an analytical reference standard for the determination of the analyte in the following:        
  • biological samples using high-performance liquid chromatography (HPLC) with fluorescence detection[3]
  • dissolved/particulate phases of wastewater and sewage sludge using liquid chromatography-electrospray-tandem mass spectrometry (LC-ESI-MS/MS)[5]        
  • whole blood samples using high-performance liquid chromatography- negative electrospray single quadrupole mass spectrometry (HPLC/ES-MS)[6]

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of eighteen perfluorinated compounds in dissolved and particulate phases of wastewater, and in sewage sludge by liquid chromatography-tandem mass spectrometry
Arvaniti SO, et al.
Analytical Methods : Advancing Methods and Applications, 6(5), 1341-1349 (2014)
Comparison of the toxicokinetic behavior of perfluorohexanoic acid (PFHxA) and nonafluorobutane-1-sulfonic acid (PFBS) in cynomolgus monkeys and rats
Chengelis PC, et al.
Reproductive Toxicology, 27(3-4), 400-406 (2009)
Validation of a screening method based on liquid chromatography coupled to high-resolution mass spectrometry for analysis of perfluoroalkylated substances in biota
Berger U and Haukas M
Journal of Chromatography A, 1801(2), 210-217 (2005)
Determination of perfluorinated carboxylic acids in biological samples by high-performance liquid chromatography
Ohya T, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 720(1-2), 1-7 (1998)
Development of a solid-phase extraction-HPLC/single quadrupole MS method for quantification of perfluorochemicals in whole blood
Karrman A, et al.
Analytical Chemistry, 77(3), 864-870 (2005)

Articles

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service