396575
Perfluoropentanoic acid
97%
Synonym(s):
Nonafluorovaleric acid, PFPeA, Nonafluoropentanoic acid, Perfluoropentanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
CF3(CF2)3COOH
CAS Number:
Molecular Weight:
264.05
Beilstein:
1800087
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.294 (lit.)
bp
140 °C (lit.)
density
1.713 g/mL at 25 °C (lit.)
SMILES string
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
InChI
1S/C5HF9O2/c6-2(7,1(15)16)3(8,9)4(10,11)5(12,13)14/h(H,15,16)
InChI key
CXZGQIAOTKWCDB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Perfluoropentanoic acid, also known as nonafluoropentanoic acid, is a monocarboxylic acid that can be utilized as a surfactant, surface protector, stain repellent, fire-fighting foam, emulsifier, and also in the production of fluoropolymers.
Application
Perfluoropentanoic acid is used as:
- An emulsifying agent for polymer dispersion, extinction foams, stain repellents, additives for paints, and coatings.
- A powerful cation exchanger in ion-pairing chromatography.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Repr. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
19F, 13C single-and two-bond 2D NMR correlations in perfluoroheptanoic acid.
Ribeiro A
Journal of Fluorine Chemistry, 83(1), 61-66 (1997)
Solubility of perfluoropentanoic acid in supercritical carbon dioxide: measurements and modeling
A Dartiguelongue, et al.
Journal of Chemical and Engineering Data, 61, 3902-3907 (2016)
Microwave spectrum of perfluoropentanoic acid
AM Pejlovas, et.al.
Chemical Physics Letters, 610, 82-85 (2014)
Aerosol synthesis and reactivity of thin oxide shell aluminum nanoparticles via fluorocarboxylic acid functional coating.
Kaplowitz D, et al.
Particle & Particle Systems Characterization, 30(10), 881-887 (2013)
Frédéric J Tessier et al.
Molecular nutrition & food research, 60(11), 2446-2456 (2016-07-10)
Nɛ -Carboxymethyl-lysine (CML) is a prominent advanced glycation end-product which is not only found in vivo but also in food. It is known that a percentage of the dietary CML (dCML) is absorbed into the circulation and only partly excreted
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service