Skip to Content
Merck
All Photos(1)

Key Documents

41610

Sigma-Aldrich

Dimethyl sulfate

puriss. p.a., ≥99.0% (GC)

Synonym(s):

Sulfuric acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2SO2
CAS Number:
Molecular Weight:
126.13
Beilstein:
635994
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.3 (vs air)

Quality Level

vapor pressure

0.7 mmHg ( 25 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

923 °F

impurities

≤0.4% free acid (as H2SO4)

ign. residue

≤0.02%

refractive index

n20/D 1.386 (lit.)
n20/D 1.387

bp

188 °C (lit.)

mp

−32 °C (lit.)

solubility

ethanol: 0.26 g/mL, clear, colorless
diethyl ether: 1.3 g/5mL, clear, colorless

density

1.333 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

SMILES string

COS(=O)(=O)OC

InChI

1S/C2H6O4S/c1-5-7(3,4)6-2/h1-2H3

InChI key

VAYGXNSJCAHWJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfate is the dimethyl ester of sulfuric acid. It undergoes hydrolysis to afford monomethyl sulfate. It is found as contaminant in fly ash and in airborne particulate matter generated during the coal combustion processes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D McGhee et al.
Proceedings of the National Academy of Sciences of the United States of America, 76(5), 2133-2137 (1979-05-01)
We have measured the effect of the histones in the nucleosome core particle on methylation of purines in nucleosome DNA by dimethyl sulfate. By using 32P terminally labeled nucleosome cores, we have examined the pattern of strand cleavage at methylated
M L Lee et al.
Science (New York, N.Y.), 207(4427), 186-188 (1980-01-11)
Dimethyl sulfate and its hydrolysis product monomethyl sulfate have been found at concentrations as high as 830 parts per million in fly ash and in airborne particulate matter from coal combustion processes. This discovery poses a new environmental problem because
Kwang Sun Ryu et al.
The journal of physical chemistry. B, 111(4), 731-739 (2007-01-26)
The physical properties of polyaniline (PAn) powder, doped by nucleophilic doping of dimethyl sulfate (DMS), were characterized, as well as its electrochemical behaviors, to investigate the possibility of a power source device adopting the PAn-DMS electrodes. It is shown that
Yogo Sakakibara et al.
Journal of the American Chemical Society, 133(22), 8396-8399 (2011-05-12)
The movement of the small ribosomal subunit (30S) relative to the large ribosomal subunit (50S) during translation is widely known, but many molecular details and roles of rRNA and proteins in this process are still undefined, especially in solution models.
Cecilia E Ström et al.
Nucleic acids research, 39(8), 3166-3175 (2010-12-25)
Base excision repair (BER) represents the most important repair pathway of endogenous DNA lesions. Initially, a base damage is recognized, excised and a DNA single-strand break (SSB) intermediate forms. The SSB is then ligated, a process that employs proteins also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service