Skip to Content
Merck
All Photos(2)

Key Documents

D100706

Sigma-Aldrich

Diethyl sulfate

98%

Synonym(s):

Ethyl sulfate, Sulfuric acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2SO2
CAS Number:
Molecular Weight:
154.18
Beilstein:
1209714
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.3 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)
2 mmHg ( 55 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.399 (lit.)

bp

208 °C (lit.)

mp

−24 °C (lit.)

density

1.177 g/mL at 25 °C (lit.)

SMILES string

CCOS(=O)(=O)OCC

InChI

1S/C4H10O4S/c1-3-7-9(5,6)8-4-2/h3-4H2,1-2H3

InChI key

DENRZWYUOJLTMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethyl sulfate can be used as a reactant for the synthesis of:
  • Biologically active compounds such as bispyrazole, pyrazolopyrimidine and pyridine containing antipyrinyl moieties.
  • N-substituted-2-styryl-4(3H)-quinazolinones.
  • Ionic liquids with pyrrolidinium, piperidinium and morpholinium cations, having potential applications as electrolytes.

It can also be used as an alkylating agent to synthesize 1-alkyl/aralkyl-2-(1-arylsufonylalkyl)benzimidazoles and an ionic liquid ethylmethylimidazole ethylsulfate ([EMIM][EtSO4].

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 1B - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 1-alkyl/aralkyl-2-(1-arylsulfonylalkyl) benzimidazoles under PTC conditions
Dubey PK, et al.
Indian Journal of Chemistry, 46B(15), 488-491 (2007)
Flow chemistry: Imidazole-based ionic liquid syntheses in micro-scale
Lowe H, et al.
Chemical Engineering Journal, 163(3), 429-437 (2010)
PEG-600-mediated, green and efficient, tandem syntheses of N-subtituted-2-styrylquinazolin-4-ones
Srinivasa RB, et al.
Green Chemistry Letters and Reviews, 6(3), 254-261 (2013)
Ionic liquids containing an alkyl sulfate group as potential electrolytes
Wu TY, et al.
Electrochimica Acta, 55(15), 4475-4482 (2010)
Jitka Caslavska et al.
Electrophoresis, 32(13), 1760-1764 (2011-05-24)
CZE coupled to sheath liquid-based electrospray ionization (ESI) and multiple-stage ion trap mass spectrometry (MS(n) ) was used for the confirmation analysis of ethyl glucuronide (EtG) and ethyl sulfate (EtS) in human serum and urine collected after intake of alcoholic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service