Skip to Content
Merck
All Photos(4)

Key Documents

36650

Sigma-Aldrich

DCC

≥99.0% (GC), for peptide synthesis

Synonym(s):

N,N′-Dicyclohexylcarbodiimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11N=C=NC6H11
CAS Number:
Molecular Weight:
206.33
Beilstein:
610662
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

DCC, puriss., ≥99.0% (GC)

grade

puriss.

Quality Level

Assay

≥99.0% (GC)

form

solid

reaction suitability

reaction type: Coupling Reactions

bp

122-124 °C/6 mmHg (lit.)

mp

32.0-37.0 °C
34-35 °C (lit.)

solubility

methylene chloride: 0.1 g/mL, clear, colorless

application(s)

peptide synthesis

SMILES string

C1CCC(CC1)N=C=NC2CCCCC2

InChI

1S/C13H22N2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h12-13H,1-10H2

InChI key

QOSSAOTZNIDXMA-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

General description

DCC is an organic compound widely used as a coupling reagent in peptide synthesis.[1][2] It is also employed as an efficient dehydrating agent for the preparation of amides, esters, nitriles, and anhydrides. Alcohols can be converted into aldehydes and ketone by Moffatt oxidation reaction in the presence of DCC and dimethyl sulfoxide (DMSO).[1][3]

Application

DCC may be used to promote the esterification of 7-((tert-butyldimethylsilyl)oxy)hepta-2,4-diyn-1-ol with propiolic acid to form 7-((tert-butyldimethylsilyl)oxy)hepta-2,4-diyn-1-yl propynoate.[4]
It may be also used to synthesize:
  • 1,3-Thiazetedine derivatives via [2+2] cycloaddition with 2-phenylethenyl- and 2-(4-nitrophenyl)ethenyl isothiocyanates.[5]
  • 1,3,5-Oxadiazine-4-thiones via [4+2] cycloaddition with benzoyl isothiocyanates.[6]
  • Sterically hindered 1,3,4-oxadiazole derivatives by reacting with (N-isocyanimino)triphenylphosphorane the presence of aromatic (or heteroaromatic) carboxylic acids.[7]

Other Notes

Coupling reagent, reagent for dehydrations etc. Modifies bovine heart mitochondrial transhydrogenase;[8][9] Inhibition of F1F0-ATPase and other proton-translocating enzymes[10]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dicyclohexylcarbodiimide (DCC)
Kvasnica M
Synlett, 2007(14), 2306-2307 (2007)
1, 3-Dicyclohexylcarbodiimide
Albert JS, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-9 (2001)
M Slebioda et al.
International journal of peptide and protein research, 35(6), 539-541 (1990-06-01)
N-acylurea, a side product in peptide synthesis from DCC, preserves its chiral integrity although peptides formed simultaneously in the same reaction are racemized to a large extent. This observation is inconsistent with the generally accepted opinion that racemization-prone O-acylisourea is
Review of synthesis of 1, 3, 4-oxadiazole derivatives.
Patel K D, et al.
Synthetic Communications, 44(13), 1859-1875 (2014)
Light-dependent chemical modification of thylakoid membrane protein with carboxyl-directed reagents.
J A Laszlo et al.
Archives of biochemistry and biophysics, 215(2), 571-581 (1982-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service