Skip to Content
Merck
All Photos(4)

Key Documents

D125407

Sigma-Aldrich

DIC

99%, for peptide synthesis

Synonym(s):

N,N′-Diisopropylcarbodiimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHN=C=NCH(CH3)2
CAS Number:
Molecular Weight:
126.20
Beilstein:
878281
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Product Name

DIC, 99%

Quality Level

Assay

99%

form

liquid

reaction suitability

reaction type: Coupling Reactions

refractive index

n20/D 1.433 (lit.)

bp

145-148 °C (lit.)

density

0.815 g/mL at 20 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)N=C=NC(C)C

InChI

1S/C7H14N2/c1-6(2)8-5-9-7(3)4/h6-7H,1-4H3

InChI key

BDNKZNFMNDZQMI-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

General description

DIC is an abbreviated form of N,N′-diisopropylcarbodiimide. It plays a role in the cyclization of N-(β-hydroxy)amides to form 2-oxazolines.

Application

Alternative to dicyclohexylcarbodiimide in peptide synthesis.
Coupling reagent for peptide syntheses.
DIC (N,N′-Diisopropylcarbodiimide) has been used in combination with 1-hydroxy-7-azabenzotriazole (HOAt) for the coupling of amino acid with N-allylglycine to form N-allylpeptide.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F

Flash Point(C)

33 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron Letters, 35, 5981-5981 (1994)
Collection of Czechoslovak Chemical Communications, 59, 691-691 (1994)
Synthesis of 2-oxazolines mediated by N,N'-diisopropylcarbodiimide.
Crosignani S, et al.
Tetrahedron Letters, 45(52), 9611-9615 (2004)
Cody L Gilleland et al.
Genetics, 201(1), 39-46 (2015-07-15)
A major goal in the study of human diseases is to assign functions to genes or genetic variants. The model organism Caenorhabditis elegans provides a powerful tool because homologs of many human genes are identifiable, and large collections of genetic
Tianyi Zhang et al.
Journal of cell science, 128(15), 2938-2950 (2015-06-21)
The v-ATPase is a fundamental eukaryotic enzyme that is central to cellular homeostasis. Although its impact on key metabolic regulators such as TORC1 is well documented, our knowledge of mechanisms that regulate v-ATPase activity is limited. Here, we report that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service