Skip to Content
Merck
All Photos(1)

Key Documents

34128

Supelco

Deoxynivalenol-13C15 solution

~25 μg/mL in acetonitrile, analytical standard

Synonym(s):

13C15 DON, Fully 13C-labelled Deoxynivalenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C15H20O6
CAS Number:
Molecular Weight:
311.21
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~25 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

mass shift

M+15

storage temp.

−20°C

SMILES string

[13CH3][13C]1=[13CH][13C@H]2O[13C@@H]3[13C@H](O)[13CH2][13C@@]([13CH3])([13C@]34[13CH2]O4)[13C@@]2([13CH2]O)[13C@H](O)[13C]1=O

InChI

1S/C15H20O6/c1-7-3-9-14(5-16,11(19)10(7)18)13(2)4-8(17)12(21-9)15(13)6-20-15/h3,8-9,11-12,16-17,19H,4-6H2,1-2H3/t8-,9-,11-,12-,13-,14-,15+/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1

InChI key

LINOMUASTDIRTM-YGEUXOLBSA-N

General description

Check out our entire range of trichothecene type mycotoxin reference materials.

Deoxynivalenol-13C15 (13C15-DON) is the 13C isotope-labelled analog of mycotoxin, Deoxynivalenol. It is a fusarium mycotoxin which belongs to trichothecene class of compounds.

Application

13C15-DON may be used as an internal standard for the identification of DON in:
  • Maize by high performance liquid chromatography (HPLC) electrospray ionization (ESI) mass spectrometry (MS/MS).
  • Cereals and cereal-based compound animal feeds by LC-MS/MS operating on selected reaction monitoring (SRM) mode.
  • Bioaerosols collected from Serpula lacrymans−damaged homes by HPLC-MS/MS in multiple reaction monitoring (MRM) mode.
  • Bread samples by reversed-phase HPLC-ESI with linear trap MS/MS.

It may also be used as an internal standard for the determination of HT-2 toxin, nivalenol and patulin in bioaerosols obtained from dairy cattle shed by HPLC-MS/MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
purity : ≥97.0% (HPLC)

Other Notes

Certan Vial

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fate of Fusarium mycotoxins in the cereal product supply chain: the deoxynivalenol (DON) case within industrial bread-making technology
Bergamini E, et al.
Food Additives and Contaminants, 27(5), 677-687 (2010)
LC-MS Based Method of Analysis for the Simultaneous Determination of four Mycotoxins in Cereals and Feed
Breidbach A, et al.
Food Additives and Contaminants, 23(11), 1187-1193 (2013)
Characterization and application of isotope-substituted (13C15)-deoxynivalenol (DON) as an internal standard for the determination of DON
Haubl G, et al.
Food Additives and Contaminants, 23(11), 1187-1193 (2006)
Recurrence of Stachybotrys chartarum during mycological and toxicological study of bioaerosols collected in a dairy cattle shed
Lanier C, et al.
Annals of Agricultural and Environmental Medicine : AAEM, 19(1), 325-334 (2012)
Airborne molds and mycotoxins in Serpula lacrymans-damaged homes
Pottier D, et al,
Atmospheric Pollution Research, 5(2), 325-334 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service