Skip to Content
Merck
All Photos(1)

Key Documents

34124

Supelco

Deoxynivalenol solution

100 μg/mL in acetonitrile, analytical standard

Synonym(s):

3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, DON, Vomitoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)([C@]34CO4)[C@@]2(CO)[C@H](O)C1=O

InChI

1S/C15H20O6/c1-7-3-9-14(5-16,11(19)10(7)18)13(2)4-8(17)12(21-9)15(13)6-20-15/h3,8-9,11-12,16-17,19H,4-6H2,1-2H3/t8-,9-,11-,12-,13-,14-,15+/m1/s1

InChI key

LINOMUASTDIRTM-QGRHZQQGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Deoxynivalenol belongs to the group of tetracyclic sesquiterpenoid mycotoxins called trichothecenes, produced by Fusarium fungi, naturally occurring in small grains and infected corn.

Check out our entire range of trichothecene type mycotoxin reference materials
Deoxynivalenol is frequently found in cereals and agricultural products including, corn, barley, oats, rye, maize, wheat, malt, bread, beer, etc. It is a mycotoxin and primarily affects the livestock industry by reducing the food intake in farm animals. The effect of DON contamination in cereals and other food samples serves as one of the factors of Kashin-Beck disease, a chronic and endemic osteoarticular disorder.

Application

Deoxynivalenol solution has been used as an analytical reference standard for the quantification of the analyte in cornmeal and wheat meal matrices using high-performance liquid chromatography with UV detection (HPLC-UV). It is used as an analytical standard in investigating its toxicity mechanism on human chondrocytes by microarray and bioinformatics analysis.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of monoclonal antibodies for the specific detection of deoxynivalenol and 15-acetyldeoxynivalenol by ELISA
Sinha.CR, et al.
Journal of Agricultural and Food Chemistry, 43, 1740-1744 (1995)
Determination of 12 type A and B trichothecenes in cereals by liquid chromatography? electrospray ionization tandem mass spectrometry
Klotzel M, et al.
Journal of Agricultural and Food Chemistry, 53, 8904-8910 (2005)
Lei Yang et al.
Toxicology letters, 321, 61-68 (2019-12-22)
T-2 toxin and deoxynivalenol (DON) are two representative mycotoxins that are commonly found in cereals and agricultural products. As T-2 toxin and DON are considered the cause of Kashin-Beck disease, a special osteoarticular disease, chondrocytes would be a vital target
Giuseppina Avantaggiato et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 42(5), 817-824 (2004-03-30)
In vitro screening of 14 adsorbent materials, including some commercial products used to detoxify Fusarium-mycotoxins, were tested in the pH range of 3-8 for deoxynivalenol (DON)- and nivalenol (NIV)-binding ability. Only activated carbon showed to be effective with binding capacities
James J Pestka
Archives of toxicology, 84(9), 663-679 (2010-08-28)
The trichothecene mycotoxin deoxynivalenol (DON) is produced in wheat, barley and corn following infestation by the fungus Fusarium in the field and during storage. Colloquially known as "vomitoxin" because of its emetic effects in pigs, DON has been associated with

Related Content

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service