Skip to Content
Merck
All Photos(1)

Key Documents

34071

Supelco

T2-Toxin solution

100 μg/mL in acetonitrile, analytical standard

Synonym(s):

Epoxytrichothecene, Fusariotoxin T 2, Mycotoxin T 2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O9
CAS Number:
Molecular Weight:
466.52
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

availability

not available in USA

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CC1=C[C@]2([H])[C@]([C@]3(C)[C@@]4(OC4)[C@]([C@H](O)[C@H]3OC(C)=O)([H])O2)(COC(C)=O)C[C@@H]1OC(CC(C)C)=O

InChI

1S/C24H34O9/c1-12(2)7-18(27)32-16-9-23(10-29-14(4)25)17(8-13(16)3)33-21-19(28)20(31-15(5)26)22(23,6)24(21)11-30-24/h8,12,16-17,19-21,28H,7,9-11H2,1-6H3/t16-,17+,19+,20+,21+,22+,23+,24-/m0/s1

InChI key

BXFOFFBJRFZBQZ-QYWOHJEZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanshen Li et al.
Journal of agricultural and food chemistry, 59(8), 3441-3453 (2011-03-23)
This review focuses on the toxicity and metabolism of T-2 toxin and analytical methods used for the determination of T-2 toxin. Among the naturally occurring trichothecenes in food and feed, T-2 toxin is a cytotoxic fungal secondary metabolite produced by
Soujanya Ratna Edupuganti et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 923-924, 98-101 (2013-03-19)
An affinity purification method that isolates T-2 toxin-specific IgY utilizing a T-2-toxin-immobilized column was developed. The T-2 toxin was covalently coupled via a carbonyldiimidazole-activated hydroxyl functional group to amine-activated sepharose beads. The affinity-purified IgY was characterized by gel electrophoresis, fast
Metabolism and pharmacokinetics of T-2 toxin and related trichothecenes.
B Yagen et al.
Drug metabolism reviews, 25(3), 281-323 (1993-01-01)
Marijana Sokolović et al.
Arhiv za higijenu rada i toksikologiju, 59(1), 43-52 (2008-04-15)
T-2 toxin is the most toxic type A trichothecene mycotoxin. It is the secondary metabolite of the Fusarium fungi, and is common in grain and animal feed. Toxic effects have been shown both in experimental animals and in livestock. It
Elin Verbrugghe et al.
Research in veterinary science, 93(3), 1139-1141 (2012-07-28)
The aim of the study was to investigate the effect of a modified glucomannan binder on the course of a Salmonella Typhimurium infection in pigs. Therefore, four pig diets were provided during 23 days: (1) free of mycotoxins, (2) containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service