Skip to Content
Merck
All Photos(1)

Key Documents

33525

Supelco

4-tert-Octylphenol-3,5-d2 monoethoxylate solution

1 μg/mL in acetone, analytical standard

Synonym(s):

2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy-3,5-d2]ethanol, 4-tert-OP1EO-D2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D2H24O2
CAS Number:
Molecular Weight:
252.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

1 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+2

storage temp.

−20°C

SMILES string

[2H]c1cc(OCCO)cc([2H])c1C(C)(C)CC(C)(C)C

InChI

1S/C16H26O2/c1-15(2,3)12-16(4,5)13-6-8-14(9-7-13)18-11-10-17/h6-9,17H,10-12H2,1-5H3/i6D,7D

InChI key

JYCQQPHGFMYQCF-QFIQSOQBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

4-tert-Octylphenol-3,5-d2 monoethoxylate solution is an isotopically labeled standard used for the analysis of mono and di-ethoxylates of nonylphenol (NP) and octylphenol (OP) in environmental samples using solid-phase micro extraction followed by gas chromatography with mass spectrometry (SPME-GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - Information refers to the main ingredient.

Flash Point(C)

-17 °C - Information refers to the main ingredient.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service