Skip to Content
Merck
All Photos(1)

Key Documents

33471

Sigma-Aldrich

1,5-Diazabicyclo[4.3.0]non-5-ene

purum, ≥98.0% (GC)

Synonym(s):

DBN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2
CAS Number:
Molecular Weight:
124.18
Beilstein:
2417
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

liquid

impurities

≤1% water

refractive index

n20/D 1.519 (lit.)
n20/D 1.521

bp

95-98 °C/7.5 mmHg (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

C1CN=C2CCCN2C1

InChI

1S/C7H12N2/c1-3-7-8-4-2-6-9(7)5-1/h1-6H2

InChI key

SGUVLZREKBPKCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 1,5-Diazabicyclo[4.3.0]non-5-ene (DBN) is an amidine base commonly used in the base-mediated eliminations, condensations, esterifications, isomerizations, carboxylations and carbonylations.
  • It is used in the preparation of supertetrahedral chalcogenide clusters and single crystals of polymer-chalcogenide composites.
  • It also acts as a catalyst for the regioselective Friedel-Crafts C-acylation of pyrroles.

Other Notes

Amidine base used for dehydrohalogenation reactions to olefins

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Oediger et al.
Synthesis, 591-591 (1972)
1, 5-Diazabicyclo [4.3. 0] non-5-ene.
Savoca, Ann C and Urgaonkar, Sameer
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Nucleophilicities and carbon basicities of DBU and DBN.
Baidya, M and Mayr, Herbert
Chemical Communications (Cambridge, England), 1792-1794 (2008)
Threading chalcogenide layers with polymer chains.
Xiong, Wei-Wei et al.
Angewandte Chemie (International Edition in English), 127(2), 556-560 (2015)
Superbase route to supertetrahedral chalcogenide clusters.
Wu, Tao et al.
Journal of the American Chemical Society, 134(8), 3619-3622 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service