Skip to Content
Merck
All Photos(1)

Key Documents

33430

Supelco

o-Dianisidine

for spectrophotometric det. of Au, NO2-, Ce(IV), for the detection of Au, Co, Cu, SCN-, V, ≥97.0%

Synonym(s):

3,3′-Dimethoxybenzidine, Fast Blue B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O2
CAS Number:
Molecular Weight:
244.29
Beilstein:
1879884
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (GC)
≥97.0%

quality

for spectrophotometric det. of Au, NO2-, Ce(IV)
for the detection of Au, Co, Cu, SCN-, V

technique(s)

UV/Vis spectroscopy: suitable

SMILES string

COc1cc(ccc1N)-c2ccc(N)c(OC)c2

InChI

1S/C14H16N2O2/c1-17-13-7-9(3-5-11(13)15)10-4-6-12(16)14(8-10)18-2/h3-8H,15-16H2,1-2H3

InChI key

JRBJSXQPQWSCCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ortho Dianisidine may be used in a novel automated method, used to measure the total antioxidant response (TAR) against potent free radical reactions.[1] It may also be used along with glucose oxidase and peroxidase in the determination of blood and urinary glucose.[2]

Warning

Probably carcinogenic.

Other Notes

Redox indicator for titration with NBS[3]

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

402.8 °F - closed cup

Flash Point(C)

206 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of lead tetraacetate, benzidine, o-dianisdine and a "film test" in investigating the periodic-acid-Schiff technic.
R E GLEGG et al.
Stain technology, 27(6), 277-305 (1952-11-01)
Determination of glucose by an improved enzymatic procedure.
Washko ME and Rice EW.
Clinical Chemistry, 7(5), 542-545 (1961)
The presence of hemoglobin within the nucleus of the embryonic chick erythroblast.
O'Brien BRA.
Experimental Cell Research, 21(1), 226-228 (1960)
H.S. Gowda et al.
Analyst, 104, 865-865 (1979)
Use of glucose oxidase, peroxidase, and O-dianisidine in determination of blood and urinary glucose.
Huggett AG and Nixon DA.
Lancet, 270 (6991), 368-370 (1957)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service