Skip to Content
Merck
All Photos(3)

Documents

A2898

Sigma-Aldrich

4-Aminobiphenyl

≥98%

Synonym(s):

4-Aminodiphenyl, 4-Biphenylamine, 4-Biphenylylamine, 4-Phenylaniline, Biphenyl-4-ylamine, NSC 7660, Xenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4NH2
CAS Number:
Molecular Weight:
169.22
Beilstein:
386533
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

autoignition temp.

842 °F

bp

191 °C/15 mmHg (lit.)

mp

52-54 °C (lit.)

SMILES string

Nc1ccc(cc1)-c2ccccc2

InChI

1S/C12H11N/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H,13H2

InChI key

DMVOXQPQNTYEKQ-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

Application

  • Induces DNA damage (carcinogen) in human bladder carcinoma cells and bladder tissue in mouse
  • Synthetic amine ligand for enrichment, depletion and one-step purification of leech proteins

Biochem/physiol Actions

Hepatic tumor initiator in experimental animal model.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dexian Dong et al.
Journal of molecular recognition : JMR, 21(3), 163-168 (2008-04-29)
Although the concept of affinity purification using synthetic ligands had been utilized for many years, there are few articles related to this research area, and they focus only on the affinity purification of specific protein by a defined library of
K L Dooley et al.
Cancer letters, 62(3), 205-209 (1992-03-15)
The tumorigenic activities of four representative heterocyclic amine food pyrolysates, 2-amino-6-methyldipyrido[1,2-a:3',2'-d]imidazole (Glu-P-1), 2-amino-3-methylimidazo[4,5-f]quinoline (IQ), 2-amino-3,8-dimethylimidazo[4,5f]quinoxaline (MeIQx), and 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP), were assessed in the neonatal male B6C3F1 mouse and were compared with that of the potent human carcinogen, 4-amino-biphenyl (4-ABP). These
Moinuddin et al.
Environmental toxicology, 29(5), 568-576 (2012-05-23)
4-Aminobiphenyl (4-ABP), an aromatic amine is a major environmental carcinogen found mainly in cigarette smoke. It has been vastly implicated in mutagenesis and cancer development. In this study, commercially available human placental DNA was exposed to 4-ABP (1.3 mM) in
Gwendoline Nauwelaers et al.
Chemical research in toxicology, 24(6), 913-925 (2011-04-05)
DNA adduct formation of the aromatic amine, 4-aminobiphenyl (4-ABP), a known human carcinogen present in tobacco smoke, and the heterocyclic aromatic amines (HAAs), 2-amino-9H-pyrido[2,3-b]indole (AαC), 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP), 2-amino-3-methylimidazo[4,5-f]quinoline (IQ), and 2-amino-3,8-dimethylmidazo[4,5-f]quinoxaline (MeIQx), potential human carcinogens, which are also present in
Dan Gu et al.
Carcinogenesis, 33(1), 124-130 (2011-11-11)
Some epidemiological investigations have revealed that frequent consumption of well-done cooked meats and tobacco smoking are risk factors for breast cancer in women. 2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) is a heterocyclic aromatic amine that is formed in well-done cooked meat, and 4-aminobiphenyl (4-ABP)

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service