Skip to Content
Merck
All Photos(1)

Key Documents

33385

Supelco

β-Endosulfan

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6Cl6O3S
CAS Number:
Molecular Weight:
406.93
Beilstein:
2950317
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[H][C@@]12COS(=O)OC[C@]1([H])C3(Cl)C(Cl)=C(Cl)C2(Cl)C3(Cl)Cl

InChI

1S/C9H6Cl6O3S/c10-5-6(11)8(13)4-2-18-19(16)17-1-3(4)7(5,12)9(8,14)15/h3-4H,1-2H2/t3-,4+,7?,8?,19-

InChI key

RDYMFSUJUZBWLH-INZWZTCPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Endosulfan is a polychlorinated compound, which is widely used as an insecticide for agricultural purposes.

Application

β-Endosulfan may be used as an analytical pesticide reference standard for the determination of the analyte in water samples, virgin olive oil, aged contaminated Ethiopian soils and human fluids by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Gas chromatographic determination of organochlorine and organophosphorus pesticides in human fluids using solid phase microextraction
Lopez FJ, et al.
Analytica Chimica Acta, 433(2), 217-226 (2001)
Acton A.Q
Benzothiepins?Advances in Research and Application: 2012 Edition: ScholarlyBrief (2012)
Development of a pressurized liquid extraction and clean-up procedure for the determination of $\alpha$-endosulfan, $\beta$-endosulfan and endosulfan sulfate in aged contaminated Ethiopian soils
Hussen A, et al.
Journal of Chromatography A, 1103(2), 202-210 (2006)
Application of single-drop microextraction and comparison with solid-phase microextraction and solid-phase extraction for the determination of $\alpha$-and $\beta$-endosulfan in water samples by gas chromatography-electron-capture detection
Lopez-Blanco MC, et al.
Journal of Chromatography A, 984(2), 245-252 (2003)
Determination of residues of endosulfan and five pyrethroid insecticides in virgin olive oil using gas chromatography with electron-capture detection.
Lentza-Rizos Ch, et al.
Journal of Chromatography A, 921(2), 297-304 (2001)

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service