Skip to Content
Merck
All Photos(1)

Key Documents

32927

Supelco

3-Acetyldeoxynivalenol

Synonym(s):

3α-Acetoxy-7α,15-dihydroxy-12,13-epoxytrichothec-9-en-8-one, 3α-Acetylvomitoxin, 3-AcDON

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22O7
CAS Number:
Molecular Weight:
338.35
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

[H][C@]12O[C@]3([H])[C@@H](C[C@@](C)([C@]34CO4)[C@@]1(CO)[C@H](O)C(=O)C(C)=C2)OC(C)=O

InChI

1S/C17H22O7/c1-8-4-11-16(6-18,13(21)12(8)20)15(3)5-10(23-9(2)19)14(24-11)17(15)7-22-17/h4,10-11,13-14,18,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17+/m1/s1

InChI key

ADFIQZBYNGPCGY-HTJQZXIKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Acetyldeoxynivalenol belongs to the class of trichothecenes, which are secondary metabolites generated by various fungi, particularly by Fusarium species.

Application

3-Acetyldeoxynivalenol may be used as an analytical reference standard for the determination of the analyte in:
  • Cereals by gas chromatography-mass spectrometry (GC-MS).
  • Milk samples by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of trichothecenes in beer by capillary gas chromatography with flame ionisation detection.
Schothorst RC and Jekel AA
Food Chemistry, 82(3), 475-479 (2003)
Michael Bretz et al.
Molecular nutrition & food research, 50(3), 251-260 (2006-03-08)
Trichothecenes are secondary metabolites produced by several fungi of the Fusarium genus during their growth period. They inhibit protein biosynthesis in eukaryotic cells resulting in numerous toxic effects such as diarrhea, vomiting, and gastro-intestinal inflammation. Considering its occurrence in food
J J Mateo et al.
Journal of chromatography. A, 918(1), 99-112 (2001-06-14)
Various analytical methods used in the analysis of type B trichothecenes (deoxynivalenol, nivalenol, 3- and 15-acetyldeoxynivalenol) in cereals were compared and optimised in this work. These methods use either GC-electron-capture detection (ECD) of trimethylsilyl, trifluoroacetyl and heptafluorobutyryl derivatives or HPLC
G S Eriksen et al.
Archiv fur Tierernahrung, 57(5), 335-345 (2003-11-19)
The absorption, metabolism and excretion of 3-acetyldeoxynivalenol (3-aDON) in pigs were studied. Pigs with a faecal microflora known to be able to de-epoxidate trichothecenes were used in the experiment. The pigs were fed a commercial diet with 3-aDON added in
A Llorens et al.
International journal of food microbiology, 94(1), 43-54 (2004-06-03)
Various species of Fusarium can produce trichothecene mycotoxins that contaminate food commodities and can represent a risk for human and animal health. In this paper, a full factorial design was applied to study the influence of incubation temperature, water activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service