Skip to Content
Merck
All Photos(2)

Key Documents

32731

Supelco

3-Amino-2-methyl-5-nitrobenzamide

VETRANAL®, analytical standard

Synonym(s):

3-ANOT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N3O3
CAS Number:
Molecular Weight:
195.18
Beilstein:
2114950
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1c(N)cc(cc1C(N)=O)[N+]([O-])=O

InChI

1S/C8H9N3O3/c1-4-6(8(10)12)2-5(11(13)14)3-7(4)9/h2-3H,9H2,1H3,(H2,10,12)

InChI key

RBLRQBGOUCRKRT-UHFFFAOYSA-N

General description

3-Amino-2-methyl-5-nitrobenzamide is a metabolite of dinitolmide which is utilized extensively as an nitroamide antiprotozoal agent in poultry farming due to its low toxicity, stability, cost-effectiveness and anticoccidial effects.[1]

Application

3-Amino-2-methyl-5-nitrobenzamide may be used as an analytical reference standard for the determination of the analyte in chicken tissues[1] and eggs[2] by accelerated solvent extraction (ASE) with a neutral alumina solid-phase extraction (SPE) column in conjunction with gas chromatography-tandem mass spectrometry (ASE-SPE-GC–MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of dinitolmide and its metabolite 3-ANOT in chicken tissues via ASE-SPE-GC-MS/MS.
Zhao X, et al.
J. Food Compos. Anal. (2018)
Development of an ASE-GC-MS/MS method for detecting dinitolmide and its metabolite 3-ANOT in eggs
Zhao X, et al.
Journal of Mass Spectrometry : Jms, 53(10), 976-985 (2018)
Gabriela A Altamirano et al.
Environmental research, 191, 110185-110185 (2020-09-19)
The plastic monomer and plasticizer bisphenol A (BPA), and the UV-filter benzophenone-3 (BP3) have been shown to have estrogenic activities that could alter mammary gland development. Our aim was to analyze whether BPA or BP3 direct exposure affects the functional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service