Skip to Content
Merck
All Photos(2)

Documents

35404

Supelco

5-Hydroxymebendazole

VETRANAL®, analytical standard

Synonym(s):

Methyl 5-[(RS)-α-hydroxybenzyl]-1H-benzimidazole-2-carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3O3
CAS Number:
Molecular Weight:
297.31
Beilstein:
4537623
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

COC(=O)Nc1nc2ccc(cc2[nH]1)C(O)c3ccccc3

InChI

1S/C16H15N3O3/c1-22-16(21)19-15-17-12-8-7-11(9-13(12)18-15)14(20)10-5-3-2-4-6-10/h2-9,14,20H,1H3,(H2,17,18,19,21)

InChI key

IIQKUGXEGMZCLE-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E G Iosifidou et al.
Drug metabolism and disposition: the biological fate of chemicals, 25(3), 317-320 (1997-03-01)
Mebendazole (MBZ) is extensively used in eel culture for treatment of Pseudodactylogyrus spp. infections. This use may lead to residues of MBZ in eel tissues. Consequently, the residue profile of MBZ in eel after treatment with the drug is of
Anthelmintic activity of methyl 5(6)-(alpha-hydroxyphenyl methyl) benzimidazole-2-carbamate, a metabolite of mebendazole.
S Gupta et al.
Indian journal of experimental biology, 25(12), 871-873 (1987-12-01)
M K Jain et al.
Indian journal of experimental biology, 30(4), 320-323 (1992-04-01)
Effect of candidate compounds 81-470 i.e. methyl [5[4-(2-pyridinyl)-1-piperazinyl]carbonyl]-1H-benzimidazole-2-yl]- carbamate and 86-162 i.e. methyl-5(6)-(alpha-hydroxyphenyl methyl) benzimidazole-2-carbamate along with reference drugs mebendazole and praziquantel on energy metabolism of C. fasciolaris recovered from rats treated with single dose of 500 mg/kg, ip was
H De Ruyck et al.
The Analyst, 126(12), 2144-2148 (2002-01-30)
A quantitative liquid chromatography-electrospray tandem mass spectrometry method for the determination of mebendazole and its hydrolysed and reduced metabolites in sheep liver has been developed and validated. The benzimidazole substances were extracted with ethyl acetate after the sample mixture had
B Oosterhuis et al.
Therapeutic drug monitoring, 6(2), 215-220 (1984-01-01)
A sensitive method is described for the simultaneous determination of mebendazole and hydroxymebendazole using flubendazole as an internal standard. The analytes were isolated with a single chloroform extraction from 1.0 ml of alkalinized plasma or cyst liquid samples. Separation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service