Skip to Content
Merck
All Photos(2)

Key Documents

E8260

Sigma-Aldrich

Ethoxyquin

≥75% (capillary GC)

Synonym(s):

1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO
CAS Number:
Molecular Weight:
217.31
Beilstein:
158223
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥75% (capillary GC)

form

liquid

density

1.03 g/mL at 20 °C (lit.)

SMILES string

CCOc1ccc2NC(C)(C)C=C(C)c2c1

InChI

1S/C14H19NO/c1-5-16-11-6-7-13-12(8-11)10(2)9-14(3,4)15-13/h6-9,15H,5H2,1-4H3

InChI key

DECIPOUIJURFOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria J Berdikova Bohne et al.
Toxicological sciences : an official journal of the Society of Toxicology, 93(1), 11-21 (2006-06-23)
The synthetic antioxidant ethoxyquin (EQ) is increasingly used in animal feeds and has been candidate for carcinogenicity testing. EQ has the potential for toxicological and adverse health effects for both fish and fish consumers through "carryover" processes. The toxicological aspects
Alina Błaszczyk et al.
Cellular & molecular biology letters, 10(1), 15-21 (2005-04-06)
In our study, we analyzed the cytotoxicity of ethoxyquin (EQ) and its two salts, ethoxyquin hydrochloride (EQ-HCL) and ethoxyquin phosphate (EQ-P). It was shown that EQ was the most cytotoxic compound (IC(50) = 0.09 mM), while the lowest cytotoxic effect
Alina Błaszczyk
Toxicology letters, 163(1), 77-83 (2005-11-04)
Ethoxyquin (1,2-dihydro-6-ethoxy-2,2,4-trimethylquinoline, EQ) is widely used in various food products and in animal feeds because of its powerful antioxidant activity. This compound was recently found to cause not only many unfavourable side-effects in animals fed with feeds containing it, but
Yoichi Aoki et al.
Journal of AOAC International, 93(1), 277-283 (2010-03-26)
An analytical method using HPLC with fluorescence detection (HPLC-FL) has been developed and applied for the survey of residue levels of ethoxyquin in a variety of food products of animal origin. HPLC was performed using a silica octadecylsilane column, butylhydroxytoluene-acetonitrile-water
J Andrew Keightley et al.
Molecular & cellular proteomics : MCP, 3(2), 167-175 (2003-12-17)
We are using a proteomic approach that combines two-dimensional electrophoresis and tandem mass spectrometry to detect and identify proteins that are differentially expressed in a cell line that is resistant to oxidative stress. The resistant cell line (OC14 cells) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service