Skip to Content
Merck
All Photos(1)

Key Documents

31116

Supelco

Nuarimol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12ClFN2O
CAS Number:
Molecular Weight:
314.74
Beilstein:
6223667
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(c1ccc(F)cc1)(c2cncnc2)c3ccccc3Cl

InChI

1S/C17H12ClFN2O/c18-16-4-2-1-3-15(16)17(22,13-9-20-11-21-10-13)12-5-7-14(19)8-6-12/h1-11,22H

InChI key

SAPGTCDSBGMXCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nuarimol may be used as a reference standard for the determination of the analyte:
  • In fresh vegetables by gas chromatography-tandem mass spectrometry (GC-MS-MS).
  • In paprika by a modified Quick Easy Cheap Effective Rugged Safe (QuEChERS) method coupled with liquid chromatography-tandem mass spectrometry (LC–MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Kostka et al.
Journal of applied toxicology : JAT, 14(5), 337-342 (1994-09-01)
It is commonly believed that in short-term tests hepatic cytochrome P-450b inducers stimulate liver enlargement and mitogenesis in the absence of overt hepatotoxic effects. In this investigation male Wistar rats received naurimol (an organochlorine pesticide) in one, three and five
D T Cooke et al.
Biochimica et biophysica acta, 1061(2), 156-162 (1991-01-30)
Oat and rye plants were treated with either tetcyclacis (an experimental plant growth regulator), nuarimol (a fungicide) or gamma-ketotriazole (an experimental herbicide). These treatments reduced shoot growth and changed the lipid composition of the shoot plasma membranes. In oat, both
B Wiadrowska et al.
Roczniki Panstwowego Zakladu Higieny, 43(3-4), 281-287 (1992-01-01)
The effect of simultaneous administration of N-diethylnitrosamine (NDEA) and Nuarimol on the activity of gamma-GTPase, G-6-Pase ans APase in the liver and serum of Wistar rats was investigated in the 2-weeks experiment. The two-stage hepato-cancerogenesis model was used with NDEA
Simón Navarro et al.
Journal of agricultural and food chemistry, 53(22), 8572-8579 (2005-10-27)
Over a 4 month brewing process, the fate of three fungicides, myclobutanil, propiconazole, and nuarimol, was studied in the spent grain, brewer wort, and final beer product. Only the residual level of myclobutanil after the mashing step was higher than
E Tyrkiel et al.
Roczniki Panstwowego Zakladu Higieny, 47(2), 151-158 (1996-01-01)
The frequency of micronuclei was assessed in polychromatic erythrocytes of bone marrow and normochromatic erythrocytes in peripheral blood of mice following exposure to fenarimol (50, 100, 200 mg/kg b.w.) and nuarimol (100, 200, 400 mg/kg b.w.) for 21 days. Mitomycin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service