1.07022
2-Propanol
≥99.5% (GC), EMPARTA® ACS reagent
Synonym(s):
sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol
About This Item
43 hPa ( 20 °C)
44 mmHg ( 25 °C)
Recommended Products
Product Name
2-Propanol, for analysis EMPARTA® ACS
grade
ACS reagent
Quality Level
vapor density
2.1 (vs air)
vapor pressure
33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)
product line
EMPLURA®
Assay
≥99.5% (GC)
form
liquid
autoignition temp.
750 °F
potency
5045 mg/kg LD50, oral (Rat)
12800 mg/kg LD50, skin (Rabbit)
expl. lim.
2.0-12.7 %, 93 °C
impurities
≤0.0001 meq/g Titrable acid
≤0.0001 meq/g Titrable base
≤0.002% Carbonyl compounds (as CO)
≤0.2% Water
evapn. residue
≤0.001%
color
clearAPHA: ≤10
refractive index
n20/D 1.377 (lit.)
pH
7 (20 °C)
bp
82 °C (lit.)
mp
−89.5 °C (lit.)
transition temp
flash point 12 °C
density
0.785 g/mL at 25 °C (lit.)
storage temp.
2-25°C
SMILES string
CC(C)O
InChI
1S/C3H8O/c1-3(2)4/h3-4H,1-2H3
InChI key
KFZMGEQAYNKOFK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Packaging
Analysis Note
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Solubility in water: conforms
Titrable acid: ≤ 0.0001 meq/g
Titrable base: ≤ 0.0001 meq/g
Carbonyl compounds (as CO): ≤ 0.002 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.2 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
53.6 °F - closed cup
Flash Point(C)
12.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.
Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.
Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.
Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service