Skip to Content
Merck
All Photos(1)

Key Documents

240486

Sigma-Aldrich

2-Methyl-2-butanol

ReagentPlus®, ≥99%

Synonym(s):

tert-Amyl alcohol, tert-Pentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C(CH3)2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1361351
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
102 °C (lit.)
vapor pressure:
12 mmHg ( 20 °C)

grade

reagent

Quality Level

vapor density

3 (vs air)

vapor pressure

12 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

819 °F

expl. lim.

9 %

dilution

(for general lab use)

refractive index

n20/D 1.405 (lit.)

bp

102 °C (lit.)

mp

−12 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCC(C)(C)O

InChI

1S/C5H12O/c1-4-5(2,3)6/h6H,4H2,1-3H3

InChI key

MSXVEPNJUHWQHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-2-butanol is a sterically hindered alcohol.

Application

2-Methyl-2-butanol may be used as a solvent in the following processes:
  • Palladium (II)-catalyzed ortho-C–H olefination of phenylacetic acids.
  • Rhodium-catalyzed direct C-H functionalization at the C7 position of N-pivaloylindoles.

It may also be used in the preparation of 2-chloro-2-methylbutane by reacting with concentrated hydrochloric acid.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

68.9 °F - closed cup

Flash Point(C)

20.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoon Ju Cho et al.
Laboratory animal research, 27(3), 197-203 (2011-10-15)
Tribromoethanol (2,2,2-tribromoethanol, TBE) is a popular injectable anesthetic agent used in mice in Korea. Our goal was to assess the risks associated with side effects (lesions) in the abdominal cavity, especially at high doses. To understand the underlying pathophysiological changes
General kinetic modeling of the selective hydrogenation of 2-methyl-3-butyn-2-ol over a commercial palladium-based catalyst.
Vernuccio S, et al.
Industrial & Engineering Chemistry Research, 54(46), 11543-11551 (2015)
Rhodium?Catalyzed Regioselective C7?Functionalization of N?Pivaloylindoles.
Xu L, et al.
Angewandte Chemie (International Edition in English), 55(1), 321-325 (2016)
?-tert-alkylation of ketones: 2-tert-pentylcyclopentanone
Organic Syntheses, 62, 95-95 (1984)
Ternary and Binary LLE Measurements for Solvent (4-Methyl-2-pentanone and 2-Methyl-2-butanol)+ Furfural+ Water between 298 and 401K.
Ma?nnisto? M, et al.
Journal of Chemical and Engineering Data, 61(2), 903-911 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service