Skip to Content
Merck
All Photos(2)

Key Documents

230987

Sigma-Aldrich

Victoria Pure Blue BO

Dye content 90 %

Synonym(s):

Basic Blue 7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40ClN3
CAS Number:
Molecular Weight:
514.14
Colour Index Number:
42595
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder, crystals or chunks

Quality Level

composition

Dye content, 90%

solubility

95% ethanol: 0.1%, blue to very deep blue

λmax

619 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CCNc1ccc(\C(c2ccc(cc2)N(CC)CC)=C3/C=C\C(C=C3)=[N+](\CC)CC)c4ccccc14

InChI

1S/C33H39N3.ClH/c1-6-34-32-24-23-31(29-13-11-12-14-30(29)32)33(25-15-19-27(20-16-25)35(7-2)8-3)26-17-21-28(22-18-26)36(9-4)10-5;/h11-24H,6-10H2,1-5H3;1H

InChI key

ROVRRJSRRSGUOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Victoria blue dyes are triaminodiphenylnaphthylmethanes, with lipophilic cations and large conjugated systems.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S M Burrow et al.
Membrane & cell biology, 14(3), 357-366 (2001-05-23)
Victoria Blue BO (VBBO) is thought to exert its photocytotoxic effects via free radical generation. Glutathione and related enzymes are associated with the protection of normal tissues against free-radical damage and have also been implicated in multiple drug resistance. It
J Morgan et al.
Photochemistry and photobiology, 67(1), 155-164 (1998-04-04)
The cationic photosensitizing triaryl methane dye Victoria Blue BO (VBBO) localizes in mitochondria and causes oxidative damage to this organelle during photodynamic therapy (PDT). Oxidative stresses from other photosensitizers induce a variety of stress proteins. The endoplasmic reticulum (ER)-based, calcium-binding
M Wainwright et al.
Journal of chemotherapy (Florence, Italy), 11(1), 61-68 (1999-03-17)
The toxicities and phototoxicities of methylene blue (MB), toluidine blue O (TBO) and Victoria blue BO (VBBO) in a murine mammary tumour cell line (EMT6-S) and a multidrug resistant (MDR) sub-line (EMT6-R) were measured and their efficacy against the resistant
J Morgan et al.
Photochemistry and photobiology, 71(6), 747-757 (2000-06-17)
The relative contribution, to cell death, of photodynamic damage to respiratory proteins (known targets of photodynamic therapy with many photosensitizers) and other cellular sites was examined. The models were a human ovarian carcinoma cell line 2008, and its mitochondrial DNA-deficient
A Viola et al.
International journal of cancer, 85(5), 733-739 (2000-03-04)
Victoria Blue BO (VB BO) is a new and promising photosensitizer currently being evaluated for photodynamic therapy (PDT). Its photochemical processes are mediated by oxygen radicals, but do not involve singlet oxygen. We used (31)P NMR spectroscopy of VB-BO sensitized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service