Skip to Content
Merck
All Photos(3)

Key Documents

227935

Sigma-Aldrich

Azure B

prepared by direct synthesis

Synonym(s):

N,N,N′-Trimethylthionin, Azure I, Methyleneazure

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN3S
CAS Number:
Molecular Weight:
305.83
Colour Index Number:
52010
Beilstein:
3922630
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

>85.0% (HPLC)

form

powder

mp

205-210 °C (dec.) (lit.)

solubility

water: 1 mg/mL, blue to very deep blue

λmax

648 nm

ε (extinction coefficient)

≥13000 at 241-247 nm
≥36000 at 287-293 nm
≥67000 at 638-644 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

C[N+](C)=C(C=C1)C=C(C1=N2)SC3=C2C=CC(NC)=C3.[Cl-]

InChI

1S/C15H15N3S.ClH/c1-16-10-4-6-12-14(8-10)19-15-9-11(18(2)3)5-7-13(15)17-12;/h4-9H,1-3H3;1H

InChI key

DNDJEIWCTMMZBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Azure B has been used in M′Fadyean stain to visualize Bacillus anthracis capsule.

Biochem/physiol Actions

Azure B is a cationic dye. It is part of the Romanowsky stain, which is used for staining of blood films. It is also used for the localization of nucleic acids and lignin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple, reliable M'Fadyean stain for visualizing the Bacillus anthracis capsule
Owen MP
Journal of Microbiological Methods, 92, 264-269 (2013)
U.S. Forest Service research note FPL (1964)
Barbara J. Bain, Imelda Bates, Mike A Laffan
Dacie and Lewis Practical Haematology (2016)
E Rosales et al.
Environmental science and pollution research international, 19(5), 1738-1746 (2011-12-14)
The aim of this work was to improve the ability of electro-Fenton technique for the remediation of wastewater contaminated with synthetic dyes using a model azo dye such as Azure B. Batch experiments were conducted to study the effects of
Luaine Bandounas et al.
New biotechnology, 30(2), 196-204 (2012-05-09)
In this study we have investigated the molecular background of the previously reported dye decolourization potential of Bacillus sp. LD003. Strain LD003 was previously isolated on Kraft lignin and was able to decolourize various lignin model dyes. Specifically Azure B

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service