Skip to Content
Merck
All Photos(1)

Key Documents

217492

Supelco

AmberChrom 50WX8 Ion Exchange Resin

hydrogen form, 50-100 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8)x
CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Amberchrom 50WX8, hydrogen form, 50-100 mesh

description

cross-linkage 8%

Quality Level

form

beads

moisture

50-58%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

50-100 mesh

capacity

1.7 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite HPR4800 Cl hydrogen form is used as stripper resin and can be regenerated by 0.25N sulphuric acid when exhausted.[1] Amberlite HPR4800 Cl hydrogen form is used as catalyst.[2] It is used in deionizing supernatant fraction samples for paper chromatography.[3]

Application

Amberlite HPR4800 Cl hydrogen form may be used in separation medium for TLC, paper chromatography and LPLC.
Strongly acidic cation exchange resin for fine chemical and pharmaceutical separations; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Other Notes

This resin has not been specially processed nor cleaned. We suggest that it be subjected to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.
Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogen generation from hydrolysis of sodium borohydride using Ni-Ru nanocomposite as catalysts.
Liu, Cheng-Hong, et al.
International Journal of Hydrogen Energy, 34, 2153-2163 (2009)
Chemical composition and ultrastructure of wild-type and white mutantAspergillus nidulans conidial walls.
Claverie-Martin, Felix, Maria R. Diaz-Torres, and Michael J. Geoghegan.
Current Microbiology, 16, 281-287 (1988)
Novel ion exchange chromatographic method using conductimetric detection.
Small, Hamish, Timothy S. Stevens, and William C. Bauman.
Analytical Chemistry, 47, 1801-1809 (1975)

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. How can I convert a cation exchange resin from the hydrogen form to the sodium form?

    1 answer
    1. To convert a cation exhcange resin from the hydrogen form to the sodium form, use two to four bed volumes of 0.5-2.0% NaCl or NaOH.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service