Skip to Content
Merck
All Photos(1)

Key Documents

217506

Supelco

AmberChrom 50WX8 Ion Exchange Resin

hydrogen form, 100-200 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8)x
CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Amberchrom 50WX8, hydrogen form, 100-200 mesh

description

cross-linkage 8%

Quality Level

form

beads

moisture

50-58%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

100-200 mesh

capacity

1.7 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite­ HPR4800 Cl hydrogen form is used as stripper resin and can be regenerated by 0.25N sulphuric acid when exhausted.[1] Amberlite HPR4800 Cl hydrogen form is used as catalyst.[2] It is used in deionizing supernatant fraction samples for paper chromatography.[3]

Application

Amberlite HPR4800 Cl hydrogen form may be used in separation medium for TLC, paper chromatography and LPLC.
Strongly acidic cation exchange resin for fine chemical and pharmaceutical separations; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Other Notes

This resin has not been specially processed nor cleaned. We suggest that it be subjected to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.
Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel ion exchange chromatographic method using conductimetric detection.
Small, Hamish, Timothy S. Stevens, and William C. Bauman.
Analytical Chemistry, 47, 1801-1809 (1975)
Chemical composition and ultrastructure of wild-type and white mutantAspergillus nidulans conidial walls.
Claverie-Martin, Felix, Maria R. Diaz-Torres, and Michael J. Geoghegan.
Current Microbiology, 16, 281-287 (1988)
Hydrogen generation from hydrolysis of sodium borohydride using Ni-Ru nanocomposite as catalysts.
Liu, Cheng-Hong, et al.
International Journal of Hydrogen Energy, 34, 2153-2163 (2009)

Questions

1–7 of 7 Questions  
  1. How can a Dowex 1x4 resin in the chloride form be converted to the acidic form?

    1 answer
    1. This product is a strongly acidic cation exchange resin in the H+ form. There is no way to convert between this and being in the chloride form, which would constitute an anion exchange resin. A resin cannot be converted from an anion exchange resin to a cation exchange resin, or vice versa. A resin in the H+ form can, however, be converted to the sodium form by flushing with two volumes of 1 N NaOH, whereby the sodium ions take the place of the hydrogen.

      Helpful?

  2. What is the meaning of 1.7 meq/mL by wetted bed volume for this resin? Do we have to consider the molecular mass of resin or the reaction mixture that we are going to purify?

    1 answer
    1. A milliequivalent is one-thousandth of an equivalent. The unit mEq is related to the number (mmol) and electrical charge of ions in solution. 1.7 mEq/mL means 1.7 millimoles of charge per milliliter of resin bed.

      Yes, you will need to consider the milliequivalents of charge on the resin and the millimoles of the material that you are trying to purify; make sure that the first is greater.

      Helpful?

  3. What is the particle size of Product 217506, Dowex® 50WX8 - hydrogen form, 100-200 mesh resin?

    1 answer
    1. This is Dowex 50Wx8 100-200 mesh, with beads measuring 75-150 microns.  This particle size is consistent in water, but the resin will shrink when exchanged to other cationic forms, and it will swell when exposed to organic solvents. You may test the swelling characteristics of a resin to be used in your application by doing the initial solvent exchanges in a graduated cylinder.

      Helpful?

  4. How do I prepare Product 217506, Dowex® 50WX8 - hydrogen form, 100-200 mesh resin, for use?

    1 answer
    1. Cation exchange resins can be prepared by initially swirling with deionized water in a flask (no stir bars, please), allowing them to settle for 15-20 minutes, and decanting the fines.  To fully activate the resin, repeat the procedure 2-3 times with 2N hydrochloric or sulfuric acid, and rinse with copious amounts of water until >pH 5; this procedure may be completed with use of a Büchner funnel.

      Helpful?

  5. Is Product 217506, Dowex® 50WX8 - hydrogen form, 100-200 mesh resin, the same as AG50W-X8?

    1 answer
    1. The AG® designation indicates that a resin has been specially cleaned and certified for analytical use.  In most cases, resin that you prepare yourself will be suitable for your application.   (® Bio-Rad Laboratories, Hercules,CA)

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  7. How can I convert a cation exchange resin from the hydrogen form to the sodium form?

    1 answer
    1. To convert a cation exhcange resin from the hydrogen form to the sodium form, use two to four bed volumes of 0.5-2.0% NaCl or NaOH.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service