07558
4-Oxoretinoic acid
analytical standard
Synonym(s):
(2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-3-oxo-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic acid, 4-Ketoretinoic acid, 4-Oxo-all-trans-retinoic acid
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥90.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
application(s)
clinical testing
format
neat
SMILES string
CC(/C=C/C1=C(C)C(CCC1(C)C)=O)=C\C=C\C(C)=C\C(O)=O
InChI
1S/C20H26O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13H,11-12H2,1-5H3,(H,22,23)/b8-6+,10-9+,14-7+,15-13+
InChI key
GGCUJPCCTQNTJF-FRCNGJHJSA-N
Recommended products
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service