Skip to Content
Merck
All Photos(2)

Documents

06737

Supelco

Green S

analytical standard

Synonym(s):

Acid Brilliant Green BS, Acid Green 50, Wool Green S

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25N2NaO7S2
CAS Number:
Molecular Weight:
576.62
Colour Index Number:
44090
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E142
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].CN(C)c1ccc(cc1)\C(=C2\C=CC(\C=C2)=[N+](/C)C)c3c(O)c(cc4cc(ccc34)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C27H26N2O7S2.Na/c1-28(2)20-9-5-17(6-10-20)25(18-7-11-21(12-8-18)29(3)4)26-23-14-13-22(37(31,32)33)15-19(23)16-24(27(26)30)38(34,35)36;/h5-16H,1-4H3,(H2,31,32,33,34,35,36);/q;+1/p-1

InChI key

WDPIZEKLJKBSOZ-UHFFFAOYSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donald R Korb et al.
Cornea, 29(4), 377-383 (2010-02-20)
The purpose of this study was to investigate the prevalence of lid wiper epitheliopathy (LWE) in patients diagnosed with dry eye disease (DED). Patients were recruited for two groups. Inclusion criteria for the DED group (n = 50) was: a
Keizo Fukushima et al.
Journal of drug targeting, 19(3), 212-218 (2010-06-15)
As a percutaneous sustained-release preparation, insulin micropiles (MPs) were prepared with biodegradable polymers poly(lactic acid) (PLA), poly(ϵ-caprolactone) (PCL) and poly(lactic-co-glycolic acid) (PLGA) as the base. The obtained PLA, PCL, and PLGA MPs of which the insulin:polymer ratio was 1:2 were
K Y Ukhanov et al.
The Journal of general physiology, 105(1), 95-116 (1995-01-01)
Several Ca-sensitive fluorescent dyes (fura-2, mag-fura-2 and Calcium Green-5N) were used to measure intracellular calcium ion concentration, Cai, accompanying light-induced excitation of Limulus ventral nerve photoreceptors. A ratiometric procedure was developed for quantification of Calcium Green-5N fluorescence. A mixture of
Masahiko Yamaguchi et al.
American journal of ophthalmology, 141(4), 669-675 (2006-03-28)
To determine whether the location of a fluorescein-stained line, the Marx line (ML), which runs along the inner eyelid, is correlated with meibomian gland function. Prospective observational case series. After applying fluorescein dye solution to the eye, the ML score
S Singh et al.
Biochemical and biophysical research communications, 195(1), 490-496 (1993-08-31)
Bio-metabolism of two permitted food colours viz., Green S and Indigo Carmine, through caecal microflora of rats was investigated. Incubation of Green S with caecal flora resulted in one coloured and two fluorescent metabolites with the corresponding Rf values of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service