Merck
  • Measurement of cytosolic Ca2+ concentration in Limulus ventral photoreceptors using fluorescent dyes.
Measurement of cytosolic Ca2+ concentration in Limulus ventral photoreceptors using fluorescent dyes.
The Journal of general physiology (1995-01-01)
K Y Ukhanov, T M Flores, H S Hsiao, P Mohapatra, C H Pitts, R Payne
ABSTRACT
Several Ca-sensitive fluorescent dyes (fura-2, mag-fura-2 and Calcium Green-5N) were used to measure intracellular calcium ion concentration, Cai, accompanying light-induced excitation of Limulus ventral nerve photoreceptors. A ratiometric procedure was developed for quantification of Calcium Green-5N fluorescence. A mixture of Calcium Green-5N and a Ca-insensitive dye, ANTS, was injected in the cell and the fluorescence intensities of both dyes were used to calculate the spatial average of Cai within the light-sensitive R lobe of the photoreceptor. In dark-adapted photoreceptors, the initial Cai was 0.40 +/- 0.22 microM (SD, n = 7) as measured with fura-2. Cai peaked in the light-sensitive R lobe at 700-900 ms after the onset of an intense measuring light step, when the spatial average of Cai within the R lobe reached 68 +/- 14 and 62 +/- 37 microM (SD, n = 5) as measured with mag-fura-2 and Calcium Green-5N, respectively. The rate of Cai rise was calculated to be approximately 350 microM/s under the measuring conditions. The resting level of Mg2+ was estimated to be 1.9 +/- 0.9 mM, calculated from mag-fura-2 measurements. To investigate the effect of adapting light on the initial Cai level in the R lobe, a 1-min step of 420 nm background light was applied before each measurement. The first significant (P < 0.05) change in the initial level of Cai occurred even at the lowest adapting light intensity, which delivered approximately 3 x 10(3) effective photons/s. The relative sensitivity of the light-adapted photoreceptors was linearly related to the relative Cai on a double log plot with slope between -4.3 and -5.3. We were unable to detect a Cai rise preceding the light-activated receptor potential. The Cai rise, measured with Calcium Green-5N, lagged 14 +/- 5 ms (SD, n = 32) behind the onset of the receptor potential at room temperature in normal ASW. In the absence of extracellular Ca2+ and at 10 degrees C, this lag increased to 44 +/- 12 ms (SD, n = 17).
Sigma-Aldrich
Lissamine Green B, Dye content 60 %
Supelco
Green S, analytical standard