Skip to Content
Merck

Skip To

02469

2-Butanone

analytical standard

Synonym(s):

Ethyl methyl ketone, MEK, Methyl ethyl ketone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
NACRES:
NA.24
PubChem Substance ID:
EC Number:
201-159-0
Beilstein/REAXYS Number:
741880
MDL number:
UNSPSC Code:
12141716
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

vapor density

2.49 (vs air)

vapor pressure

71 mmHg ( 20 °C)

assay

≥99.9% (GC)

autoignition temp.

960 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

10.1 %

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.379

bp

80 °C (lit.)

mp

−87 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(CC)=O

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

General description

2-Butanone also known as methyl ethyl ketone is a colorless fairly volatile liquid with a pleasant pungent odor. It is primarily used as a solvent, for making other chemicals, and for production of wax from petroleum. It is derived from refluxing methyl acetoacetate and dilute sulphuric acid. It finds application in manufacturing plastic, textile, paints, pharmaceuticals and cosmetics.

Application

2-Butanone may have been used as reference standard in solid-phase microextraction gas-chromatography (SPME-GC) for quantification of volatile compounds from dry-cured hams.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
676926W217018PHR1207
grade

analytical standard

grade

ACS reagent

grade

FG, Halal, Kosher

grade

certified reference material, pharmaceutical secondary standard

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

application(s)

-

application(s)

flavors and fragrances

application(s)

pharmaceutical (small molecule)

assay

≥99.9% (GC)

assay

≥99.0%

assay

≥99.5% (GC)

assay

-

format

neat

format

-

format

-

format

neat

mp

−87 °C (lit.)

mp

−87 °C (lit.)

mp

−87 °C (lit.)

mp

−87 °C (lit.)


Still not finding the right product?

Explore all of our products under 2-Butanone


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

30.2 °F - closed cup

flash_point_c

-1 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Characterization of French and Spanish dry-cured hams: influence of the volatiles from the muscles and the subcutaneous fat quantified by SPME-GC.
Sanchez-Pe?a CM
Meat Science, 69(4), 635-645 (2005)
Cooper's Toxic Exposures Desk Reference with CD-ROM, 377-377 (1996)
Chao Long et al.
Journal of hazardous materials, 203-204, 251-256 (2011-12-30)
A novel hypercrosslinked polymeric adsorbent (HY-1) with high surface area and specific bimodal pore size distribution in the regions of micropore (0.5-2.0 nm) and meso-macropore (30-70 nm) was prepared. Adsorption properties of benzene and methyl ethyl ketone (MEK) vapors onto



Global Trade Item Number

SKUGTIN
M2705-1G04061825590592
02469-5ML04061837239731

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters