Merck
  • Removal of benzene and methyl ethyl ketone vapor: comparison of hypercrosslinked polymeric adsorbent with activated carbon.
Removal of benzene and methyl ethyl ketone vapor: comparison of hypercrosslinked polymeric adsorbent with activated carbon.
Journal of hazardous materials (2011-12-30)
Chao Long, Ying Li, Weihua Yu, Aimin Li
ABSTRACT
A novel hypercrosslinked polymeric adsorbent (HY-1) with high surface area and specific bimodal pore size distribution in the regions of micropore (0.5-2.0 nm) and meso-macropore (30-70 nm) was prepared. Adsorption properties of benzene and methyl ethyl ketone (MEK) vapors onto HY-1 were investigated and compared with a commercial microporous activated carbon (m-GAC). The equilibrium adsorption data showed that the adsorption capacities of benzene and MEK on HY-1 were larger than those of m-GAC at the higher relative pressure. The Dubinin-Radushkevich (D-R) equation was found to fit the experimental data well. The isosteric enthalpy of adsorption for benzene and MEK were calculated. The m-GAC exhibited much higher values of ΔH(st) for the VOCs than HY-1 at the whole loading studied, which can lead to significant temperature rises during the adsorption step. The results of dynamic experiments revealed that HY-1 had a good dynamic adsorption capacity with a longer breakthrough time and shorter length of mass transfer zone due to its specific bimodal property. Therefore, HY-1 will be a particularly efficient and competitive adsorbent for VOCs recovery, especially at medium-high concentrations.
Product Description
Supelco
2-Butanone, analytical standard
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
2-Butanone, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, ReagentPlus®, ≥99%