Skip to Content
Merck
All Photos(1)

Documents

02300

Sigma-Aldrich

D-(+)-Malic acid

unnatural form, ≥97.0% (T)

Synonym(s):

(R)-(+)-2-Hydroxysuccinic acid, D-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723540
EC Number:
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

optical activity

[α]20/D +28.0±2°, c = 5.5% in pyridine

quality

unnatural form

mp

98-102 °C (lit.)

SMILES string

O[C@H](CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m1/s1

InChI key

BJEPYKJPYRNKOW-UWTATZPHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(+)-Malic acid can be used:
  • As a starting material for the enantioselective total synthesis of (−)-erinapyrone B.
  • As a chiral organocatalyst in the synthesis of α-aminophosphonates from various aldehydes, aniline, and diethyl phosphite.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concise Total Synthesis of (-)-Erinapyrone B from D-(+)-Malic Acid
Samala R, et al.
Synthetic Communications, 44(4), 500-506 (2014)
R Kaminsky et al.
Tropical medicine & international health : TM & IH, 1(2), 255-263 (1996-04-01)
The unique features of purine salvage systems of pathogenic haemoflagellates render them selectively susceptible to the cytotoxic effects of purine analogues. A series of acyclic nucleoside phosphonates were evaluated for activity against pathogenic haemoflagellates in vitro. One of the phosphonylmethoxyalkylpurines
One pot green synthesis of α-aminophosphonates with D-Malic acid as an organocatalyst
AIP Conference Proceedings, 1860(1), 020057-020057 (2017)
K Leckel et al.
Clinical and experimental immunology, 134(2), 238-245 (2003-11-18)
Immunosuppression correlates with the development and recurrence of cancer. Mycophenolate mofetil (MMF) has been shown to reduce adhesion molecule expression and leucocyte recruitment into the donor organ. We have hypothesized that MMF might also prevent receptor-dependent tumour dissemination. Therefore, we
C Orskov et al.
FEBS letters, 247(2), 193-196 (1989-04-24)
We developed specific, C-terminal radioimmunoassays for three proglucagon (PG) fragments: PG 151-158, PG 151-160 and PG 126-159 (glucagon-like peptide-2 (GLP-2] in order to determine the exact C-terminal sequence of the newly isolated GLP-2 in man and pig. The antigens and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service