Skip to Content
Merck
All Photos(5)

Documents

O4126

Sigma-Aldrich

Oxaloacetic acid

≥97% (HPLC)

Synonym(s):

2-Oxosuccinic acid, Ketosuccinic acid, Oxalacetic acid, Oxobutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2COCOOH
CAS Number:
Molecular Weight:
132.07
Beilstein:
1705475
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

powder

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to light yellow

storage temp.

−20°C

SMILES string

OC(=O)CC(=O)C(O)=O

InChI

1S/C4H4O5/c5-2(4(8)9)1-3(6)7/h1H2,(H,6,7)(H,8,9)

InChI key

KHPXUQMNIQBQEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxaloacetic acid is a dicarboxylic acid. It is an intermediate in the citric acid cycle. It is highly soluble in water and is present ubiquitously. It is produced in the mitochondria by the action of pyruvate carboxylase on pyruvate. Breakdown products of oxaloacetate includes malate, pyruvate and aspartic acid.

Application

Oxaloacetic acid has been used as a substrate for measuring citrate synthase activity in cybrids and neuroblastoma cells. It has also been used for measuring malate dehydrogenase reactivation.

Biochem/physiol Actions

Oxaloacetic acid being an intermediate in the tri carboxylic cycle is central to metabolism. It is part of gluconeogenesis pathway. Mutation in pyruvate carboxylase leads to decreased production of oxaloacetate. It inhibits succinate dehydrogenase and is a key regulator of mitochondrial metabolism.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxaloacetic acid mediates ADP-dependent inhibition of mitochondrial complex IIdriven respiration
Fink BD, et al.
The Journal of Biological Chemistry, 293(51), 1993219941-1993219941 (2018)
Synergistic coordination of polyethylene glycol with ClpB/DnaKJE bichaperone for refolding of heat-denatured malate dehydrogenase
Nian R, et al.
Biotechnology Progress, 25(4), 10781085-10781085 (2009)
Oxaloacetic acid supplementation as a mimic of calorie restriction
Cash A
Open Longevity Science, 3, 2227-2227 (2009)
Improved mitochondrial and methylglyoxal-related metabolisms support hyperproliferation induced by 50 Hz magnetic field in neuroblastoma cells
Falone S, et al.
Journal of Cellular Physiology, 231(9), 20142025-20142025 (2016)
MitoTALEN: a general approach to reduce mutant mtDNA loads and restore oxidative phosphorylation function in mitochondrial diseases
Hashimoto M, et al.
Molecular Therapy, 23(10), 15921599-15921599 (2015)

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

See All

Protocols

Spectrophotometric assay evaluates malic dehydrogenase activity using bovine heart enzyme with critical histidine residue at active site.

Spectrophotometric assay evaluates malic dehydrogenase activity using bovine heart enzyme with critical histidine residue at active site.

Spectrophotometric assay evaluates malic dehydrogenase activity using bovine heart enzyme with critical histidine residue at active site.

Spectrophotometric assay evaluates malic dehydrogenase activity using bovine heart enzyme with critical histidine residue at active site.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service