Skip to Content
Merck
All Photos(1)

Key Documents

00285

Sigma-Aldrich

1-Butylpyridinium bromide

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14BrN
CAS Number:
Molecular Weight:
216.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

≥99.0%

impurities

≤0.2% water

SMILES string

[Br-].CCCC[n+]1ccccc1

InChI

1S/C9H14N.BrH/c1-2-3-7-10-8-5-4-6-9-10;/h4-6,8-9H,2-3,7H2,1H3;1H/q+1;/p-1

InChI key

KVBQNFMTEUEOCD-UHFFFAOYSA-M

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
BR88615BR88635BR88610
packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

manufacturer/tradename

BRAND 88630

manufacturer/tradename

BRAND 88615

manufacturer/tradename

BRAND 88635

manufacturer/tradename

BRAND 88610

I.D.

4 mm

I.D.

2 mm

I.D.

4 mm

I.D.

-

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service