Skip to Content
Merck
All Photos(1)

Key Documents

BR88610

BRAND® stopcock one-way with nozzles

for tube I.D. 3 mm

Synonym(s):

PTFE, Stopcocks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NB.13
Pricing and availability is not currently available.

material

PTFE

packaging

pack of 1 ea

manufacturer/tradename

BRAND 88610

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The BRAND® one-way stopcock with nozzles is constructed from high-quality PTFE, offering outstanding chemical resistance for various laboratory applications. This stopcock features a valve lever made of polypropylene (PP) for easy operation and control. It is equipped with integrated hose nozzles, facilitating efficient fluid transfer and connection to hoses. The robust design ensures reliable performance, making it suitable for handling aggressive chemicals and solvents.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service