Skip to Content
Merck
All Photos(2)

Key Documents

11534378910

Roche

Tetramethyl-Rhodamine-5-dUTP

Synonym(s):

Rhodamine-5-dUTP tetralithium salt, Tetramethylrhodamine-5(6)-(5-[3-carboxamidoallyl]-2′-deoxyuridine 5′-triphosphate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H40N5O18P3
Molecular Weight:
935.66
UNSPSC Code:
41116100

grade

Molecular Biology

Quality Level

Assay

97.7% (HPLC)

form

solution

mol wt

990.5

packaging

pkg of 25 μL (25 nmol; 1mM)

manufacturer/tradename

Roche

concentration

1.0 mmol/L (Tetramethylrhodamine-5-dUTP (Abs. 551 nm))

color

clear red

mp

~0 °C

solubility

water: miscible

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethylrhodamine is bound to deoxyuridine triphosphate via an amide linkage.

Application

Tetramethyl-Rhodamine-5-dUTP has been used in the labelling of DNA fragments and in the synthesis of fluorescence in situ hybridization (FISH) probes.

Quality

Typical analysis: At least 85% Tetramethyl-Rhodamine-5-dUTP (HPLC, area%).

Analysis Note

Absorption: Absorption spectra available in the files.

Other Notes

For life science research only. Not for use in diagnostic procedures. For some in situ experiments, it is useful to label DNA with two fluorescent "tags," tetramethylrhodamine (red) and fluorescein (yellow).

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyan Tao et al.
Comparative cytogenetics, 15(4), 375-392 (2021-11-23)
Kengyiliahirsuta (Keng, 1959) J. L. Yang, C. Yen et B. R. Baum, 1992, a perennial hexaploidy species, is a wild relative species to wheat with great potential for wheat improvement and domestication. The genome structure and cross-species homoeology of K.hirsuta
Developmental stage influences chromosome segregation patterns and arrangement in the extremely polyploid, giant bacterium Epulopiscium sp. type B
Hutchison E, et al.
Molecular Microbiology, 107(1), 68-80 (2018)
Robert Hasterok et al.
Genetics, 173(1), 349-362 (2006-02-21)
As part of an initiative to develop Brachypodium distachyon as a genomic "bridge" species between rice and the temperate cereals and grasses, a BAC library has been constructed for the two diploid (2n = 2x = 10) genotypes, ABR1 and
rDNA cytogenetics and some structural variability in an Avena barbata Pott ex Link? A. sativa subsp. nuda (L.) Gillet et Magne amphiploid after 5-azaC treatment.
Florek M and Kosina R
Genetic Resources and Crop Evolution, 64(7), 1723-1741 (2017)
Dominika Idziak et al.
PloS one, 9(3), e93503-e93503 (2014-03-29)
Paleogenomic studies based on bioinformatic analyses of DNA sequences have enabled unprecedented insight into the evolution of grass genomes. They have revealed that nested chromosome fusions played an important role in the divergence of modern grasses. Nowadays, studies on karyotype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service