Skip to Content
Merck
All Photos(10)

Key Documents

ZIQ7000T0C

Milli-Q

Milli-Q® IQ 7000 Ultrapure Water Purification System

greener alternative

output: type 1 water (18.2 MΩ·cm), the most advanced Milli-Q® ultrapure (Type 1) water purification system that is intelligent, intuitive, and green.

Synonym(s):

Milli-Q® IQ 7000 Ultrapure Water Purification System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104206
NACRES:
NB.85

AC/DC input

100 V / 230 V AC, 50-60 Hz

Quality Level

product line

Milli-Q® IQ

packaging

pkg of 1 unit

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

parameter

16.0 kg operating weight (35.3 lb)
300 L/day max. usage

system H × W × D

49.8 cm (19.6 in.) × 26.5 cm (10.4 in.) × 35 cm (13.8 in.)

input

feed water nature pure water

output

product water quality: type 1 water (18.2 MΩ·cm)

conductivity

0.055 μS/cm at 25 °C (Product Water)

application(s)

PFAS testing

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

The Milli-Q® IQ 7000 Ultrapure Water System features advanced technologies, hydraulic design, and software capabilities. It has been built in a compact, ergonomic and intelligent way to deliver superior quality ultrapure water. The system comprises two separate and distinct components. The Milli-Q® ultrapure water production unit offers flexible installation and more simplified maintenance. The new Q-POD® point-of-delivery unit provides final polishing adapted to application needs by removing specific types of contaminants. A large touch screen allows for intuitive interaction and displays essential data for complete user reassurance. Up to four new Q-POD® components can be used with each production unit at different locations within the same laboratory, or in adjacent laboratories.
For more information on this product please contact our Technical Service Team

Application

This advanced Milli-Q® ultrapure water system is intelligent, intuitive, and green. It is used for sensitive analytical techniques: HPLC, UHPLC, LC-MS, IC, and other types of chromatography, as well as for AAS, ICP-MS, and in studies about nanoparticles. It is also used to prepare buffers and reagents in cell biology, immunostaining, molecular biology and biochemistry.

Features and Benefits

MyMilli-Q Digital Services

Be fully supported by MyMilli-Q Remote Care online monitoring and service capability, for your lab’s maximum productivity. Immediate and customizable notifications of system alerts and alarms 24/7, by e-mail and SMS.Real-time system information, performance, water quality data and more on your computer or mobile device.Rapid, online diagnostics – and even remote repairs possible – by our service team.Full data traceability and effortless data access and retrieval capabilities facilitate audit preparation and lab accreditation.Online management of your service and consumable contracts. MyMilli-Q Digital Services Be fully supported by MyMilli-Q Remote Care online monitoring and service capability, for your lab’s maximum productivity. Immediate and customizable notifications of system alerts and alarms 24/7, by e-mail and SMS. Real-time system information, performance, water quality data and more on your computer or mobile device.Rapid, online diagnostics – and even remote repairs possible – by our service team. Full data traceability and effortless data access and retrieval capabilities facilitate audit preparation and lab accreditation.Online management of your service and consumable contracts.
Water Purification System

  • Simple & intuitive dispensing: Obtaining ultrapure water from the redesigned Q-POD® dispenser has never been so easy, precise or effortless with the new "wheel" dispense. Access multiple dispensing functions that are useful for day-to-day work in the lab.
  • Intuitive touch screen interactions: Like a smartphone, the Q-POD® dispenser′s large, interactive touch screen allows you to go anywhere in just a few clicks.
  • Optimized lab space: Protect your working space from clutter. The system is designed for a neat installation and clean lab benches, free of loose tubing and cables. You only need to interact with the Q-POD® dispenser - the rest of the compact system can easily be integrated into the lab environment. For labs needing more than one ultrapure water point-of-use, up to four Q-POD® dispensers can be connected to one production unit.
  • Superior ultrapure water - consistently - to the trace level: The combination of Jetpore® and IQnano resins allow you to fully rely on the system′s water purity for your critical experiments. And with A10® TOC on-board monitoring, you′re spared from uncertainty or misinterpreting results - and also from having to repeat time-consuming analyses.
  • Advanced traceability: Advanced data tracking, such as Dispensing Events ensures a history of all dispense reports and Daily Quality Measures provides water quality traceability over time. Complete History is a global archive of all system data when more in-depth analysis is required. Traceability is also ensured through the e-Sure tags on system POD-Paks and cartridges.
  • Quality & Compliance: The system is intended to produce ultrapure water that meets or exceeds requirements as described by the ASTM®, ISO 3696, the CLSI norms, European Pharmacopoeia and US Pharmacopoeia.
  • Product Water TOC: < 2 ppb (in the appropriate operating conditions, otherwise typically < 5 ppb).
  • Product water particulates: No particles with a size > 0.22 mm (with MILLIPAK® filter).
  • Product water instant delivery rate: Up to 2 L/min.

Other Notes

Greener Alternative Product: The system has been evaluated on 7 dimensions that have a critical impact on Global Warming. Officially awarded the Greener Alternative Product label, we certify that the Milli-Q® IQ 7000 system uses less plastic and less electricity, and is completely mercury-free. View the Design for Sustainability (DfS) scorecard for the Milli-Q® IQ 7000 system.

Components required but not included:

  • A specific country box with the user manual in the local language and a power cord adapted to the local electrical network needs to be ordered in addition to this catalog number.
  • The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
  • This catalog number does not include consumables, accessories, and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!

Legal Information

A10 is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
JETPORE is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
MyMilli-Q is a trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Time-resolved analysis of dissolution phenomena in photoelectrochemistry-A case study of WO3 photocorrosion
Knoppel J, et al.
Electrochemical Communications, 96, 53-56 (2018)
Visualizing Potential-Induced Pitting Corrosion of Ultrathin Single-Crystalline IrO2 (110) Films on RuO2 (110)/Ru (0001) under Electrochemical Water Splitting Conditions
Weber T, et al.
ChemCatChem, 12(3), 855-866 (2020)
Separation and Characterization of New Forced Degradation Products of Dasatinib in Tablet Dosage Formulation Using LC-MS and Stability-Indicating HPLC Methods
Lakka N S, et al.
Chromatographia, 1-16 (2020)
27933586
Bolla P K, et al.
Journal of drug delivery science and technology, 49, 6-13 (2019)
Yu Kang et al.
Journal of chromatography. A, 1585, 138-143 (2018-12-07)
Both fully (98%) and partially (88%) hydrolyzed poly(vinyl alcohol) (PVA) are characterized by aqueous gel permeation chromatography (GPC) coupled with multi-angle laser light scattering. Polymer adsorption on hydrophilic GPC columns is observed for partially hydrolyzed PVA but not for fully

Articles

Development of an HPLC-UV method to determine Arbutin and Hydroquinone in whitening serum using a Chromolith® HighResolution C18 100x2mm HPLC column.

Development of an HPLC-UV method to determine Arbutin and Hydroquinone in whitening serum using a Chromolith® HighResolution C18 100x2mm HPLC column.

Development of an HPLC-UV method to determine Arbutin and Hydroquinone in whitening serum using a Chromolith® HighResolution C18 100x2mm HPLC column.

Development of an HPLC-UV method to determine Arbutin and Hydroquinone in whitening serum using a Chromolith® HighResolution C18 100x2mm HPLC column.

Related Content

Discover an ultrapure lab water system designed with you in mind. Intelligent, intuitive and connected, the Milli-Q® IQ 7000 system will simplify the way you work in the lab.

Discover an ultrapure lab water system designed with you in mind. Intelligent, intuitive and connected, the Milli-Q® IQ 7000 system will simplify the way you work in the lab.

Discover an ultrapure lab water system designed with you in mind. Intelligent, intuitive and connected, the Milli-Q® IQ 7000 system will simplify the way you work in the lab.

Discover an ultrapure lab water system designed with you in mind. Intelligent, intuitive and connected, the Milli-Q® IQ 7000 system will simplify the way you work in the lab.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service