Skip to Content
Merck
All Photos(6)

Key Documents

IPAKKIT00

Millipore

Milli-Q® Polishing Kit for Purified Water

new
greener alternative

For Milli-Q® IQ/EQ 7000 and EQ 7008/16 ultrapure water systems fed with purified water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212

Quality Level

packaging

units of 1

manufacturer/tradename

Milli-Q®

storage condition

room temperature

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

cartridge H

39.5 cm (15.5 in.)

input

feed water nature purified water

compatibility

for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IQ 7000

greener alternative category

shipped in

ambient

General description

This Milli-Q® kit contains IPAK Meta® and IPAK Quanta® polishing cartridges for purified water, designed to work as a pair to produce ultrapure (Type 1) water for sensitive laboratory applications.
The cartridges contain a combination of Jetpore® mixed-bed ion-exchange resin and innovative IQnano ion-exchange media to remove trace ions from purified feed water, as well as a high-grade synthetic activated carbon, which targets trace organic contaminants.
We continue to demonstrate our commitment to a sustainable future by exploring ways to improve existing products in our portfolio. We have reviewed our purification cartridges to identify key opportunities to minimize impact without compromising on the product performance. This product has benefited from an optimization of the resin mix, the packaging and the end-of-life management. Therefore, it is identified as a greener alternative product in the category reengineered. View the DfS scorecard for the new Milli-Q® polishing kit for pure water.

Application

  • For Milli-Q® IQ/EQ 7000 and EQ 7008/16 ultrapure water systems when fed with purified water.
  • Removes trace ions and organic contaminants to deliver ultrapure (Type 1) water for sensitive applications.

Features and Benefits

  • e-Sure tags enable RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with one Drain Cap per cartridge, to remove excess water and reduce the weight of exhausted cartridges at disposal.
  • 78% of the parts suppliers for the cartridges included in this kit are ISO 14001 environmental management system-certified.

Legal Information

IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
JETPORE is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.

Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.

Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.

Reduce your lab’s consumption of water, energy, plastics and chemicals. Eco-friendly Milli-Q® lab water purification systems and digital services can support your lab’s sustainability goals.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service