Skip to Content
Merck
All Photos(1)

Key Documents

WP6111560

Millipore

Chemical Duty Pump, 115 V/60 Hz

Synonym(s):

Laboratory vacuum pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105101
eCl@ss:
32012303
NACRES:
NB.24

AC/DC input

115 V, 60 Hz

shipped in

ambient

General description

  • Chemical Duty Pump is ideal for use in life science, environmental, and industrial applications.
  • Features a chemically-resistant head and diaphragm ideal for use with corrosive chemicals and solvents.
  • Withstands solvent vapor and moisture
  • Includes a 70 cm of 1/4 in. tubing and a Millex FA50 filter for in-line moisture protection.
  • Easy-to-adjust vacuum and pressure regulators with individual gauges.
  • Portable design for easy movement between workstations.

Application

Vacuum source for filter holders, manifolds, and devices, including Simplicity® Filtration System, SNAP id® 2.0 System, Multiscreen® HTS Filtration Manifold, and Stericup® filters.

Principle

Vacuum

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Millex is a trademark of Merck KGaA, Darmstadt, Germany
SIMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.

An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.

An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.

An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.

Protocols

HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note.

HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note.

Related Content

Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.

Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.

Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.

Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service