Skip to Content
Merck
All Photos(1)

Key Documents

OP10L

Sigma-Aldrich

Anti-c-Myc (Ab-1) Mouse mAb (9E10)

lyophilized, clone 9E10, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9E10, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

rat (weakly), human, mouse (weakly)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)
mouse ... Myc(17869)
rat ... Myc(24577)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with SP2/0 mouse myeloma cells. Recognizes the ~64-67 kDa (apparent MW) c-Myc protein.
Recognizes a ~60-67 kDa c-Myc protein in HL60 cells and lung carcinoma tissue.
This Anti-c-Myc (Ab-1) Mouse mAb (9E10) is validated for use in FC, Frozen Sections, Immunoblotting, IF, IP, Chromatin IP, Paraffin Sections for the detection of c-Myc (Ab-1).

Immunogen

Epitope: within amino acids 410-419
Human
a synthetic peptide (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) corresponding to amino acids 408-439 of human c-Myc

Application

Flow Cytometry (20 µg/ml or use Cat. No. OP10F)

Frozen Sections (2-10 µg/ml)

Immunoblotting (1-5 µg/ml, see application references)

Immunofluorescence (1-5 µg/ml or use Cat. No. OP10F)

Immunoprecipitation (1 µg/sample)

Chromatin Immunoprecipitation (5 µg/ml, see application references)

Paraffin Sections (not recommended)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a volatile buffer, 100 µg BSA.

Reconstitution

Reconstitute the lyophilized antibody with sterile PBS, pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 µg/ml. Lyophilized antibodies should be resuspended at 4°C with occasional gentle mixing for at least 2 h.

Analysis Note

Negative Control
HT1080 cells
Positive Control
HL-60 cells or lung carcinoma

Other Notes

LeGouy, E., et al. 1987. In Nuclear Oncogenes, Cold Spring Harbor Laboratory, 144.
Cole, M.D. 1986. Ann. Rev. Gen.20, 361.
Nisen, P.D., et al. 1986. Cancer Res.46, 6217.
Nau, M.M., et al. 1985. Nature318, 69.
Persson, H., et al. 1984. Science225, 687.
Alitalo, K., et al. 1983. Proc. Natl. Acad. Sci. USA80, 1707.
The c-Myc protein is extremely labile. c-Myc degradation can be as a result of freeze/thaw cycles, thus, we recommend using fresh lysates with a cocktail of protease inhibitors. c-Myc is often destroyed during the processing of paraffin sections and can therefore be difficult to detect in this application. This antibody recognizes the c-Myc protein and its cleavage products. The sequence of c-Myc predicts a ~45 kDa protein, but c-Myc migrates under reducing conditions as a ~64-67 kDa band. This antibody will not detect v-Myc but may react weakly to rodent c-Myc when used at high concentrations (10 µg/ml). An extra ~34-40 kDa band may be detected in a immunoblot. May also be used in immunofluorescence as well as the detection and purification of recombinant proteins tagged with the Myc epitope sequence EQKLISEEDL (see application references). The immunogen, c-Myc (Peptide-1) is also available for competition studies (Cat. No. PP06). Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazutaka Iijima et al.
Oncology letters, 22(1), 546-546 (2021-08-03)
Six-transmembrane epithelial antigen of the prostate 1 (STEAP1) has emerged as an ideal target in cancer therapeutics. However, the functions of STEAP1 in liver cancer remain unexplored. The current study aimed to characterize the biological roles of STEAP1 in liver
Yusuke Kamihara et al.
Oncotarget, 7(39), 64330-64341 (2016-09-08)
Deregulated iron metabolism underlies the pathogenesis of many human cancers. Recently, low expression of ferroportin, which is the only identified non-heme iron exporter, has been associated with significantly reduced overall survival in multiple myeloma (MM); however, the altered iron metabolism
P Loflin et al.
FEBS letters, 509(2), 267-271 (2001-12-14)
Differentiation-dependent expression of the Na(+)/glucose cotransporter (SGLT1) is accompanied by a large, cAMP-dependent increase in stability of its mRNA. Stabilization is mediated by protein binding to a critical uridine-rich element (URE) in its 3' untranslated region. In the present study
Esther Camp et al.
Stem cells (Dayton, Ohio), 27(9), 2081-2091 (2009-06-23)
Nanog is involved in controlling pluripotency and differentiation of stem cells in vitro. However, its function in vivo has been studied only in mouse embryos and various reports suggest that Nanog may not be required for the regulation of differentiation.
Chuan-Fen Wu et al.
Developmental biology, 308(1), 169-186 (2007-06-15)
Tumorhead (TH) regulates neural plate cell proliferation during Xenopus early development, and gain or loss of function prevents neural differentiation. TH shuttles between the nuclear and cytoplasmic/cortical cell compartments in embryonic cells. In this study, we show that subcellular distribution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service