Skip to Content
Merck
All Photos(1)

Documents

MAB88916

Sigma-Aldrich

Anti-Fibronectin Frag Antibody, cell attachment fragment, clone 3E3

clone 3E3, Chemicon®, from mouse

Synonym(s):

Anti-CIG, Anti-ED-B, Anti-FINC, Anti-FN, Anti-FNZ, Anti-GFND, Anti-GFND2, Anti-LETS, Anti-MSF, Anti-SMDCF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)

Specificity

The antibody reacts with human fibronectin and with a peptic fragment (11.5 kD) from human fibronectin (Pierschbacker et al., 1981) comprimising the cell attachment site (Pierschbacker et al., 1981; Pierschabacker et al., 1983, 1984; Hayman et al., 1985; Ruoslahti et al., 1986; Hynes, 1985). It inhibits the attachment of normal rat kidney cells to human fironectin coated plastic surface (Pierschbacker et al., 1981). Binding of the antibody to fibronectin cannot be inhibited by cell adhesion peptides from fibronectin having the sequence Arg-Gly-Asp of Gly-Arg-Gly-Asp-Ser (Pierschabacker et al., 1984; Hayman et al., 1985; Ruoslahti et al., 1986).

Immunogen

Epitope: cell attachment fragment
Purified human-fibronectin from amnion liquid.

Application

Anti-Fibronectin Frag Antibody, cell attachment fragment, clone 3E3 is an antibody against Fibronectin Frag for use in ELISA, WB, IC, IH.
Immunocytochemistry: (5-20 μg/mL) Immunohistochemistry: (5-20 μg/mL) (Not tested on paraffin sections) Immunoblotting: (5-20 μg-mL)

ELISA

Inactivation or inhibition studies in cell culture.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oncogenic targeting of BRM drives malignancy through C/EBP?-dependent induction of ?5 integrin.
Damiano, L; Stewart, KM; Cohet, N; Mouw, JK; Lakins, JN; Debnath, J; Reisman et al.
Oncogene null
Manuel L B Palacio et al.
Journal of biomedical materials research. Part A, 100(1), 18-25 (2011-10-06)
Conformational changes of fibronectin (Fn) deposited on poly(methyl methacrylate) and poly(acrylic acid) block copolymers with identical chemical compositions were detected using an antibody-functionalized atomic force microscope (AFM) tip. Based on the antibody-protein adhesive force maps and phase imaging, it was
Tomoko Nagase et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 238(5), 1118-1130 (2009-04-23)
In routine culture, human embryonic stem (hES) cells are maintained on either feeder cells or special culture substrates such as Matrigel. However, to expand hES cells for clinical applications, it is desirable to minimize animal-derived materials in the culture for
Wei Liu et al.
Cell and tissue research, 360(2), 245-262 (2015-02-11)
Cochlear micromechanics and frequency tuning depend on the macromolecular organization of the basilar membrane (BM), which is still unclear in man. Novel techniques in cochlear implantation (CI) motivate further analyses of the BM. Normal cochleae from patients undergoing removal of
Bryan A McLendon et al.
Biology, 10(6) (2021-07-03)
Cells respond to extracellular mechanical forces through the assembly of integrin adhesion complexes (IACs) that provide a scaffold through which cells sense and transduce responses to those forces. IACs are composed of transmembrane integrin receptors that bind to extracellular matrix

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service